Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301212

Sigma-Aldrich

Anti-CYP7A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CP7A, CYP7, CYPVII

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

1.4 mg/mL

technique(s)

immunohistochemistry: 1:25- 1:100

isotype

IgG

accession no.

NP_000771.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP7A1(1581)

Specificity

The antibody detects endogenous levels of total CYP7A1 protein.

Immunogen

Synthetic peptide corresponding to a region derived from internal residues of human cytochrome P450, family 7, subfamily A, polypeptide 1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Hee Kim et al.
PloS one, 18(6), e0287146-e0287146 (2023-06-13)
Macrophage inhibitory cytokine 1 (MIC-1), which is overproduced in various human cancers and associated with cachexia, acts on the hypothalamus to suppress appetite and reduce body weight. We investigated the mechanisms through which MIC-1 affects bile acid metabolism and gallstone
Eun-Ji Lee et al.
International journal of molecular medicine, 41(3), 1715-1723 (2017-12-30)
Gallstone disease is one of the most prevalent and costly gastrointestinal disorders worldwide. Gallstones are formed in the biliary system by cholesterol secretions in bile, which result from excess cholesterol, a deficiency in bile salts or a combination of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service