Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300883

Sigma-Aldrich

Anti-NeuN antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FLJ56884, FLJ58356, FOX3, HRNBP3, RBFOX3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$434.00

$434.00

PRICE WITHOUT NATIONAL TAXES

Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$434.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$434.00

PRICE WITHOUT NATIONAL TAXES

Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

human, mouse, rat

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:2000 (Cell Lysate)

isotype

IgG

accession no.

NP_001076044.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBFOX3(146713)

General description

NeuN (neuronal nuclei), also known as RBFOX3 (RNA binding protein, fox-1 homolog 3), is encoded by the gene mapped to human chromosome 17q25.3. The encoded protein belongs to the RBFOX family of proteins. NeuN is present in nuclei and perinuclear cytoplasm of most of the neurons in the central nervous system (CNS) of mammals.

Specificity

The antibody detects endogenous level of total NeuN protein.

Immunogen

Recombinant protein of human NeuN.

Application

Anti-NeuN antibody produced in rabbit has been used in Immunohistochemical staining.

Biochem/physiol Actions

NeuN (neuronal nuclei)/RBFOX3 (RNA binding protein, fox-1 homolog 3) is a RNA splicing factor. It controls neuronal splicing pathway. NeuN is used as a potential marker of postmitotic neurons in studies of neuronal differentiation and in the evaluation of neuronal status both in norm and pathology. Mutation in the gene is associated with risk of rolandic epilepsy. Studies have shown that in presence of neurological disorders, the protein is mislocalized.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 1.0mg/mL in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electroacupuncture promotes neural stem cell proliferation and neurogenesis in the dentate gyrus of rats following stroke via upregulation of Notch1 expression.
Zhao J, et al.
Molecular Medicine Reports, 12(5), 6911-6917 (2015)
RBFOX and PTBP1 proteins regulate the alternative splicing of micro-exons in human brain transcripts.
Li Y I, et al.
Genome Research, 25(1), 1-13 (2015)
Effect of transplantation of olfactory ensheathing cell conditioned medium induced bone marrow stromal cells on rats with spinal cord injury.
Feng L, et al.
Molecular Medicine Reports, 16(2), 1661-1668 (2017)
ETV2 mediates endothelial transdifferentiation of glioblastoma.
Zhao C, et al.
Signal transduction and targeted therapy, 3(1), 4-4 (2018)
Chengjian Zhao et al.
Signal transduction and targeted therapy, 3, 4-4 (2018-03-13)
Glioblastoma multiforme (GBM) is characterized by extensive endothelial hyperplasia. Recent studies suggest that a subpopulation of endothelial cells originates via vasculogenesis by the transdifferentiation of GBM tumor cells into endothelial cells (endo-transdifferentiation). The molecular mechanism underlying this process remains poorly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service