Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300830

Sigma-Aldrich

Anti-UCN antibody produced in rabbit

affinity isolated antibody

Synonym(s):

UI, UROC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

concentration

0.7 mg/mL

technique(s)

immunohistochemistry: 1:25- 1:100

isotype

IgG

accession no.

NP_003344.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCN(7349)

General description

Urocortin (UCN) belongs to the hypothalamic hormone corticotrophin-releasing factor (CRF) family. UCN has 45% homology to CRF. UCN is expressed in brain, placenta, lymphocytes and heart. In human chromosome, the gene UCN is localized on 2p23.3.

Specificity

The antibody detects endogenous levels of total UCN protein.

Immunogen

Synthetic peptide corresponding to residues near the C terminal of human urocortin

Application

Anti-UCN antibody produced in rabbit has been used in immunohistochemical analysis.

Biochem/physiol Actions

Urocortin (UCN1) is a key protein involved in regulating hypothalamic-pituitary-adrenal axis and modulating stress responses. Ucn1 is associated with both G-protein coupled CRF receptors and CRF binding proteins. UCN increases the heart rate, cardiac output, and coronary blood flow according to the dosage. UCN protects cardiac myocytes from cell death due to stress. Ucn upregulates p42/p44 mitogen activated protein kinase (MAPK) signalling pathway and protects the heart from reperfusion injury. High levels of Ucn mRNA have been observed in the Edinger−Westphal (EW) nucleus, associated with alcohol and drug addiction. Ucn delays the gastric emptying and reduces the food intake and thus regulates the body weight. Ucn enhances anxiogenic-like or stress-like effect. Ucn causes degranulation of mast cells and increases vascular permeability in response to stress. Ucn is found to be associated with stress induced atopic dermatitis or psoriasis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W J Giardino et al.
Translational psychiatry, 7(1), e1021-e1021 (2017-02-01)
Midbrain neurons of the centrally projecting Edinger-Westphal nucleus (EWcp) are activated by alcohol, and enriched with stress-responsive neuropeptide modulators (including the paralog of corticotropin-releasing factor, urocortin-1). Evidence suggests that EWcp neurons promote behavioral processes for alcohol-seeking and consumption, but a
Urocortin reduces food intake and gastric emptying in lean and ob/ob obese mice
Asakawa A, et al.
Gastroenterology, 116(6), 1287-1292 (1999)
Control of chronic excessive alcohol drinking by genetic manipulation of the Edinger-Westphal nucleus urocortin-1 neuropeptide system
Giardino WJ, et al.
Translational Psychiatry, 7(1), e1021-e1021 (2017)
Urocortin protects against ischemic and reperfusion injury via a MAPK-dependent pathway
Brar BK, et al.
The Journal of Biological Chemistry, 275(12), 8508-8514 (2000)
Urocortin protects the heart from reperfusion injury via upregulation of p42/p44 MAPK signaling pathway
Schulman D, et al.
American Journal of Physiology. Heart and Circulatory Physiology, 52(4), H1481-H1488 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service