Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300470

Sigma-Aldrich

Anti-CTNNB1 (Ab-33) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CTNNB antibody produced in rabbit, Anti-DKFZp686D02253 antibody produced in rabbit, Anti-FLJ25606 antibody produced in rabbit, Anti-FLJ37923 antibody produced in rabbit, Anti-catenin (cadherin-associated protein), beta 1, 88kDa antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~92 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(L-D-S-G-I)

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)

General description

Catenin beta-1 (CTNNB1) encoding β-catenin, belongs to armadillo-repeat family. In human chromosome, the gene CTNNB1 is localized on 3p22.1.

Immunogen

Peptide sequence around aa. 31-35 (L-D-S-G-I), according to the protein CTNNB1.

Biochem/physiol Actions

Catenin beta-1 (CTNNB1) is a signalling molecule in the nucleus and anchors actin cytoskeleton to cadherin-based adherens junctions. Phosphorylation causes dissociation of β-catenin from E-cadherin and degradation by proteasome pathway. β-catenin is a key molecule in wingless/integrated (wnt) signalling. β-catenin forms a complex with kindlin 2 and T-cell factor 4 (TCF4) to regulate wnt target gene expression. Mutations in adenomatous polyposis coli (APC) and CTNNB1 gene affects β-catenin intracellular localization and stability, contributing to aberrant expression of wnt/β-catenin in malignant melanoma. β-catenin is a major oncoprotein, which downregulates death associated protein kinase 2 (Dapk2) and inhibits anoikis of cancer cells. β-catenin expression is suppressed and its downstream gene transcription function is regulated by a tumor suppressor protein fragile histidine triad (Fhit). High expression of mucin 1 (MUC1), translocates β-catenin out of the nucleus and inhibits β-catenin regulated cell proliferation. Somatic mutations of CTNNB1 in glycogen synthase kinase 3β−casein kinase 1 (GSK-3β−CK1) phosphorylation site results in loss of phosphorylation of β-catenin. This causes aberrant wnt-signalling and leads to adrenocortical adenomas and adrenocortical carcinoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Involved in the regulation of cell adhesion and in signal transduction through the Wnt pathway.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compartmentalized CDK2 is connected with SHP-1 and beta-catenin and regulates insulin internalization
Fiset A, et al.
Cellular Signalling, 23(5), 911-919 (2011)
Pregnancy, primary aldosteronism, and adrenal CTNNB1 mutations
Teo AED, et al.
The New England Journal of Medicine, 373(15), 1429-1436 (2015)
The tumor suppressor HINT1 regulates MITF and beta-catenin transcriptional activity in melanoma cells
Genovese G, et al.
Cell Cycle, 11(11), 2206-2215 (2012)
Down-regulation of death-associated protein kinase-2 is required for beta-catenin-induced anoikis resistance of malignant epithelial cells
Li H, et al.
The Journal of Biological Chemistry, 284(4), 2012-2022 (2009)
Molecular genetic analysis of malignant melanomas for aberrations of the WNT signalling pathway genes CTNNB1, APC, ICAT and BTRC
Reifenberger J, et al.
International Journal of Cancer. Journal International Du Cancer, 100(5), 549-556 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service