Skip to Content
Merck
All Photos(4)

Documents

SAB4200815

Sigma-Aldrich

Anti-VEGF165 antibody, Mouse monoclonal

clone VG-20, purified from hybridoma cell culture

Synonym(s):

VEGF-A, VPF, Vascular endothelial growth factor A, Vascular permeability factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

VG-20, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using recombinant human VEGFA protein expressed in HEK-293

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VEGF165(7422)

General description

VEGFA - Vascular Endothelial Growth Factor A, also known as or Vascular permeability factor (VPF), is a major signaling growth factor member of the VEGF family, sub-group of the Platelet-Derived Growth Factor (PDFG) proteins, which is characterized by its conserved cystine-knot structure.1 VEGF expression is up regulated in hypoxia and in many types of cancers. Furthermore, VEGF receptors (VEGFR1 and VEGFR2) are also overexpressed in several tumors and studies have shown that high levels of VEGF correlates with poor prognosis.1-8 Inhibition of VEGF mediated angiogenesis signaling pathways have potential in angiogenesis-related disease therapy, this can be achieved by blocking either the circulating VEGF using Anti-VEGF antibodies such as Bevacizumab and Aflibercept5 or by VEGFR-2 inhibition using small molecules such as Gamabufotalin (CS-6) and Arenobufagin.6-8

Specificity

Monoclonal Anti-VEGF165 specifically recognizes human VEGFA isoform VEGF165 and does not recognize VEGF121 isoform.

Immunogen

Recombinant VEGF165 protein

Application

The antibody is recommended to use in various immunological techniques, including Immunoblot (~21 kDa). Detection of the VEGF165 band by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

VEGF proteins are involved in angiogenesis and vasculogenesis, they predominantly activate proliferation and survival of endothelial cells and migration in non-endothelial cells.1VEGFA have several isoforms due to alternative splicing mainly at exons 6-8. The VEGF exon 8 splicing isoforms are divided into two groups, the “pro-angiogenic” VEGFxxxa isoforms considered to be activators of proliferation, survival and angiogenesis and the VEGFxxxb isoforms that are considered as “anti-angiogenic”.2

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavia Plastino et al.
Experimental eye research, 206, 108518-108518 (2021-02-28)
Echinomycin (EKN), an inhibitor of hypoxia-inducible factor (HIF)-1 DNA-binding activity, has been implied as a possible therapeutic agent in ischemic diseases. Here, we assess EKN in hypoxia-driven responses in vitro using human primary adult retinal pigment epithelium cells (aRPE) and
Pooja Yadav et al.
iScience, 26(6), 106804-106804 (2023-05-26)
Alternative splicing of vascular endothelial growth factor A (VEGFA) generates numerous isoforms with unique roles in tumor angiogenesis, and investigating the underlying mechanism during hypoxia necessitates diligent pursuance. Our research systematically demonstrated that the splicing factor SRSF2 causes the inclusion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service