Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200676

Sigma-Aldrich

Monoclonal Anti-SPAG11 antibody produced in mouse

clone 10B4, purified from hybridoma cell culture

Synonym(s):

Anti-Spag11, Anti-Spag11a/b, Bin1b, sperm associated antigen 11A/B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10B4, monoclonal

form

buffered aqueous solution

mol wt

monomer ~7 kDa
~34 kDa (multimer)

species reactivity

rat, mouse

concentration

~1 mg/mL

technique(s)

immunoblotting: 4-8 μg/mL using whole extracts of HEK-293 cells overexpressing rat SPAG11
immunohistochemistry: 5-10 μg/mL using paraffin-embedded rat caput epididimys sections

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-SPAG11 (mouse IgG1 isotype) is derived from the hybridoma 10B4 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. SPAG11 is expressed specifically in epithelial cells in the middle part of the caput region. It exists in four isoforms such as A, D, E and G.
SPAG11A (sperm associated antigen 11A) belongs to the β defensin family. The protein is mainly expressed in the epididymis. The gene is mapped to human chromosome 8.

Specificity

Monoclonal Anti-SPAG11 recognizes mouse and rat SPAG11.

Immunogen

a recombinant rat SPAG11

Application

Monoclonal Anti-SPAG11 antibody produced in mouse may be used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

SPAG11 is involved in promoting sperm motility due to its capability to bind the sperm head in different regions of the epididymis. It exhibits anti-microbial activity by suppressing colony growth of Escherichia coli and other Gram-negative bacteria.
SPAG11A (sperm associated antigen 11A) is a secretory protein with possible involvement in sperm maturation. It also controls inflammatory responses in the epididymis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing or storage in “frost-free” freezer is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dwi A Pujianto et al.
Reproductive biology and endocrinology : RB&E, 11, 59-59 (2013-07-03)
Epididymal sperm maturation occurs via interactions between sperm and proteins secreted by the epididymal epithelium. Although this is an important process, the genes that encode the involved proteins remain largely uncharacterized. Previous studies have demonstrated that the genes involved in
Multimerin-2 maintains vascular stability and permeability
Pellicani R, et al.
Matrix Biology, 87, 11-25 (2020)
Identification, cloning and functional characterization of novel sperm associated antigen 11 (SPAG11) isoforms in the rat
Yenugu S, et al.
Reproductive Biology and Endocrinology, 4(1), 1-14 (2006)
Z Fei et al.
Genes and immunity, 13(6), 445-451 (2012-04-27)
The mBin1b is a beta-defensin gene identified in the mouse epididymis. In the current report, its expression pattern and antibacterial activities were characterized, and a transgenic (TG) mouse model was developed in which mBin1b was exclusively overexpressed by up to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service