Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200429

Sigma-Aldrich

Anti-PIWIL4 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-HIWI2, Anti-MIWI2, Anti-Piwi-like 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~95 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using lysates of HEK-293Tcells overexpressing human PIWIL4.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIWIL4(143689)

General description

P-element Induced WImpy testis (PIWI)-like 4 (PIWIL4) protein belongs to the PIWI subfamily. This protein is localized in tumorous colon tissues. PIWIL4 gene is located on the human chromosome at 11q21.

Specificity

Anti-PIWIL4 recognizes human PIWIL4.

Immunogen

peptide corresponding to an internal region of human PIWIL4, conjugated to KLH. The corresponding sequence differs by a single amino acid in mouse and rat.

Application

Anti-PIWIL4 antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

P-element Induced WImpy testis (PIWI)-like 4 (PIWIL4) protein promotes tumorigenesis and cell invasion. It is expressed in several cancers like colon and breast cancer. This protein enhances cell growth and blocks apoptosis in cervical cancer cells. PIWIL4 protein is also involved in the chromatin-modifying pathway of human somatic cells. Mutation in the PIWIL4 gene is associated with breast cancer.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Su et al.
FEBS letters, 586(9), 1356-1362 (2012-04-10)
Recently, PIWIL4 has been identified as a functional protein involved in tumorigenesis. Cervical cancer is the second most prevalent form of cancer worldwide. The relationship between PIWIL4 and cervical cancer is still unknown. Here, we found that PIWIL4 is up-regulated
Zifeng Wang et al.
The Journal of biological chemistry, 291(20), 10646-10658 (2016-03-10)
P-element-induced wimpy testis (PIWI) proteins bind to PIWI-interacting RNAs and play key roles in the biogenesis and functions of PIWI-interacting RNAs. It has been reported that PIWI proteins are essential for stem cell self-renewal and germline development in diverse organisms
Kaipeng Xie et al.
Journal of ovarian research, 12(1), 1-1 (2019-01-06)
Ovarian cancer remains the most fatal gynecologic malignancy worldwide due to delayed diagnosis as well as recurrence and drug resistance. Thus, the development of new tumor-related molecules with high sensitivity and specificity to replace or supplement existing tools is urgently
Keiki Sugimoto et al.
Biochemical and biophysical research communications, 359(3), 497-502 (2007-06-05)
The field of epigenetics has made progress by the identification of the small RNA-mediated epigenetic modification. However, little is known about the key proteins. Here, we report that the human PIWI-like family is a candidate protein that is involved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service