Pricing and availability is not currently available.
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~120 kDa
species reactivity
human
concentration
~1.0 mg/mL
technique(s)
immunoprecipitation (IP): 5-10 μg using lysates of human WiDr cells. indirect immunofluorescence: 1-2 μg/mL using human HeLa cells. western blot: 0.5-1.0 μg/mL using whole extracts of human WiDr cells
Junction mediating and regulatory protein (JMY) is identified as a transcriptional co-activator of p53 and is a multifunctional actin nucleation factor. JMY contains Wiskott-Aldrich syndrome protein (WASp)-homology 2 (WH2) domains in its C-terminus. The JMY gene is mapped to human chromosome location 5q14.1.
Specificity
Anti-JMY recognizes human JMY.
Immunogen
synthetic peptide corresponding to the C-terminal region of human JMY, conjugated to KLH.
Application
Anti-JMY antibody produced in rabbit may be used in:
immunoblotting
immunoprecipitation
immunofluorescence.
Biochem/physiol Actions
Junction mediating and regulatory protein (JMY) acts as a nucleation promoting factor by activating actin-related proteins-2/3 (ARP2/3), creating branched filament networks, and also by assembling unbranched filaments directly through a Spire-like mechanism. Overexpression of JMY increases motility while its loss slows cell migration. DNA damage causes JMY to accumulate in the nucleus, where it enhances p53-dependent transcription of pro-apoptotic genes. The activities of JMY as a p53 co-factor or as an actin nucleation factor are regulated by its subcellular localization.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Storage and Stability
For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.
Disclaimer
Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
p53-cofactor JMY is a multifunctional actin nucleation factor
Zuchero JB, et al.
Nature Cell Biology, 11(4), 451-459 (2009)
A transcription co-factor integrates cell adhesion and motility with the p53 response
Coutts AS, et al.
Proceedings of the National Academy of Sciences, 106(47), 19872-19877 (2009)
Identification of differentially expressed genes in uveal melanoma using suppressive subtractive hybridization
Landreville S, et al.
Molecular Vision, 17, 1324-1324 (2011)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.