Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200352

Sigma-Aldrich

Anti-DLC1 (C-terminal) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-ARHGAP7, Anti-Deleted in Liver Cancer 1, Anti-FLJ21120, Anti-HP, Anti-STARD12, Anti-p122-RhoGAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~200 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using lysates of HEK-293T cells overexpressing human DLC1.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DLC1(10395)

General description

Deleted in liver cancer 1 (DLC1) is a candidate tumor suppressor gene, originally obtained from human hepatocellular carcinoma (HCC). It is mapped to the human chromosome location 8p21-22. The DLC1 gene is majorly expressed in normal human tissues. It contains three major functional domains, sterile α-motif (SAM), Rho guanosine triphosphatase-activating protein (RhoGAP), and steroidogenic acute regulatory-related lipid transfer (START) domains. The N-terminal region of DLC1 localizes to focal adhesions by binding to Src Homology 2 (SH2) domains on tensins.

Specificity

Anti-DLC1(C-terminal) specifically recognizes human DLC1.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminal of human DLC1, conjugated to KLH. The antibody is affinity-purified using the immunizing peptide immobilized on agarose.

Application

Anti-DLC1 (C-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Deleted in liver cancer 1 (DLC1) interacts with tensin2, a focal adhesion protein, localized at the end of stress fibers, which plays key roles in cytoskeletal organization and cell signaling. Mutations in the focal adhesion targeting (FAT) region of DLC1 reduces its expression and function. The focal adhesion localization of DLC1 is critical for cell motility and morphology. It is also critical for the tumor suppressor activity of DLC1 and under expressed in human hepatocellular carcinoma (HCC) and a variety of cancer types.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deleted in liver cancer 1 expression and localization in hepatocellular carcinoma tissue sections
Wolosz D, et al.
Oncology Letters, 8(2), 785-788 (2014)
Mutations in the focal adhesion targeting region of deleted in liver cancer-1 attenuate their expression and function
Liao YC, et al.
Cancer Research, 68(19), 7718-7722 (2008)
Focal adhesion-localization of START-GAP1/DLC1 is essential for cell motility and morphology
Kawai K, et al.
Genes Cells, 14(2), 227-241 (2009)
Role of DLC1 tumor suppressor gene and MYC oncogene in pathogenesis of human hepatocellular carcinoma: potential prospects for combined targeted therapeutics
Zimonjic DB and Popescu NC
International Journal of Oncology, 41(2), 393-406 (2012)
Deleted in liver cancer-1 (DLC-1): a tumor suppressor not just for liver
Liao YC and Lo SH
The International Journal of Biochemistry & Cell Biology, 40(5), 843-847 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service