Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200284

Sigma-Aldrich

Anti-Neurocalcin-δ antibody produced in rabbit

enhanced validation

~0.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-NCALD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~22 kDa

species reactivity

human, mouse, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~0.5 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using mouse brain extract (S1 fraction) and lysates of HEK-293T cells over-expressing human neurocalcin-δ.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCALD(83988)
mouse ... Ncald(52589)
rat ... Ncald(553106)

General description

The neurocalcin-δ (NCALD) gene is mapped to human chromosome 8q22.3. The encoded protein belongs to a large family of neuronal calcium (Ca2+) sensor (NCS) proteins and is a member of visinin-like protein (VILIP) subfamily. Neurocalcin-δ is also one of the EF-hand-containing Ca2+-binding proteins. It is predominantly expressed in the Purkinje cells of the cerebellum and is also present in the olfactory epithelium.

Specificity

Anti-Neurocalcin-d specifically recognizes human and mouse neurocalcin-d.

Immunogen

synthetic peptide corresponding to a sequence located near the N-terminus of human neurocalcin-δ, conjugated to KLH. The corresponding sequence is identical in mouse and rat neurocalcin-δ.

Application

Anti-Neurocalcin-δ antibody produced in rabbit is suitable for immunoblotting.

Biochem/physiol Actions

Neurocalcin-δ (VILIP-3) is a neuronal calcium (Ca2+) sensor. It binds cooperatively to three Ca2+ ions. Neurocalcin-δ influences cyclic guanosine monophosphate (cGMP) signaling in the retina, by acting as a regulator of the retinal guanylate cyclase (retGC1)/ photoreceptor membrane guanylyl cyclase 1 (ROSGC1). Neurocalcin-δ interacts with retGC1 in a calcium (Ca2+) dependent manner. It colocalizes with retGC1 in neurons in the inner plexiform layer of the retina, in amacrine and ganglion cells where it may influence synaptic signaling processes. Neurocalcin-δ has been shown to interact with actin, clathrin, and α- and β-tubulins, indicating that it may be involved with clathrin-coated vesicle traffic. It interacts and activates the olfactory guanylate cyclase ONE-GC, suggesting that it may function in regulating the odorant signaling pathway.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 1% BSA as stabilizer and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Ping Chen et al.
Taiwanese journal of obstetrics & gynecology, 56(6), 843-846 (2017-12-16)
We present prenatal diagnosis of an interstitial 8q22.2-q23.3 deletion associated with bilateral cleft lip and palate and intrauterine growth restriction (IUGR) on fetal ultrasound. A 29-year-old, primigravid woman underwent elective amniocentesis at 17 weeks of gestation because of anxiety. Amniocentesis
Teresa Duda et al.
FEBS letters, 583(8), 1327-1330 (2009-03-25)
In a subset of olfactory epithelium the odorant receptor guanylate cyclase, ONE-GC, is a central transduction component of the cyclic GMP signaling pathway. The odorant binds to the extracellular domain and activates its intracellular catalytic domain to generate the odorant
The calcium sensor protein visinin-like protein-1 modulates the surface expression and agonist sensitivity of the alpha4beta2 nicotinic acetylcholine receptor
Lin L, et al.
The Journal of biological chemistry, 41872-41878 null
Venkateswar Venkataraman et al.
Biochemistry, 42(19), 5640-5648 (2003-05-14)
Rod outer segment membrane guanylate cyclase (ROS-GC) transduction system is a central component of the Ca(2+)-sensitive phototransduction machinery. The system is composed of two parts: Ca(2+) sensor guanylate cyclase activating protein (GCAP) and ROS-GC. GCAP senses Ca(2+) impulses and inhibits
Lenka Ivings et al.
The Biochemical journal, 363(Pt 3), 599-608 (2002-04-20)
The neuronal calcium sensors are a family of EF-hand-containing Ca(2+)-binding proteins expressed predominantly in retinal photoreceptors and neurons. One of the family members is neurocalcin delta, the function of which is unknown. As an approach to elucidating the protein interactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service