Skip to Content
Merck
All Photos(3)

Documents

SAB4100235

Sigma-Aldrich

Monoclonal Anti-SAMHD1 antibody produced in mouse

clone SAMHD1-A200, culture supernatant

Synonym(s):

AntiAGS5, AntiDCIP, AntiHD domain containing 1, AntiHDDC1, AntiMOP-5, AntiOTTHUMP00000030889, AntiSAM domain and HD domain 1, AntiSAM domain- and HD domain-containing protein 1, AntiSBBI88

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SAMHD1-A200, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 72 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SAMHD1(25939)

General description

SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1 (SAMHD1) is a deoxynucleoside triphosphate triphosphohydrolase with sterile alpha motif (SAM) and a dGTP-regulated dNTP hydrolase domain (HD). SAMHD1 gene is mapped to human chromosome 20q11.23.

Immunogen

SAMHD1 purified recombinant protein

Biochem/physiol Actions

SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1 (SAMHD1) is a key enzyme, which controls the levels of dinucleotide triphosphates. It also acts as an immune response factor cum restriction factor for many virues including the human immunodeficiency virus (HIV) hepatitis B virus (HBV) and herpes simplex virus 1 (HSV-1). SAMHD1 interferon-stimulated gene (ISG) also regulates the human cytomegalovirus and papillomavirus life cycles. Mutation in the SAMHD1 leads to imbalance in dinucleotide levels and is implicated in Aicardi‐Goutières syndrome (AGS) Mutation in SAMHD1 along with Three Prime Repair Exonuclease 1 (TREX1) and Stimulator of interferon genes (STING) genes is also correlated to familial chilblain lupus (FCL).

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SAMHD1-deficient fibroblasts from Aicardi-Goutieres Syndrome patients can escape senescence and accumulate mutations
Franzolin E, et al.
Faseb Journal, 34(1), 631-647 (2020)
Autosomal dominant inheritance of a heterozygous mutation in SAMHD1 causing familial chilblain lupus
Ravenscroft JC, et al.
American Journal of Medical Genetics. Part A, 155(1), 235-237 (2011)
SAMHD1 Regulates Human Papillomavirus 16-Induced Cell Proliferation and Viral Replication during Differentiation of Keratinocytes
James CD, et al.
mSphere, 4(4), e00448-e00419 (2019)
Microarray CGH analysis of hematological patients with del (20q)
Wu C, et al.
International Journal of Hematology, 102(5), 617-625 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service