Skip to Content
Merck
All Photos(1)

Key Documents

SAB3700792

Sigma-Aldrich

Anti-Ferret IgG (γ-chain specific) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

ferret

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Ferret IgM coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Goat Serum, Ferret IgG and Ferret Serum. No reaction was observed against Ferret IgM or Ferret IgA. Specificity was confirmed by ELISA at less than 1% cross reactivity against other Ferret heavy or light chain isotypes.

Immunogen

Ferret IgG gamma heavy chain

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.1 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronja Mathiesen et al.
Acta veterinaria Scandinavica, 60(1), 36-36 (2018-06-20)
The significance of maternal immunoglobulin G (IgG) for the resistance against a number of infections affecting the health of young mink offspring is not known. Here, we present a validated immunoassay for quantification of mink IgG in serum and milk

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service