Skip to Content
Merck
All Photos(1)

Key Documents

SAB3700041

Sigma-Aldrich

Anti-Bovine IgM (μ-chain specific) antibody produced in rabbit

whole antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$310.00

$310.00

PRICE WITHOUT NATIONAL TAXES

Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
2 ML
$310.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$310.00

PRICE WITHOUT NATIONAL TAXES

Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

bovine

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

General description

During the B-cell development, IgM (immunoglobulin M) is the first expressed antibody. It is present as a monomer on the surface of naive B cells. Antigenic stimulation and upon maturation, the monomeric IgM units (190kDa), are linked by disulfide bonds to form a pentameric immunoglobulin structure. It also but rarely forms a hexameric structure. The pentameric IgM is linked by a disulfide bond, a polypeptide chain and a J-chain. The μ -chain consists of five domains- VH, Cμ1, Cμ2, Cμ3 and Cμ4.

Specificity

This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against Bovine IgM and Bovine Serum. No reaction was observed against Bovine IgG F(ab′)2 or Bovine IgG F(c).

Immunogen

Bovine IgM mu heavy chain

Biochem/physiol Actions

The J-chain of the multimeric IgM (immunoglobulin M) structure is responsible for secretion at the mucosal surfaces. The pentameric form of IgM, has a greater affinity towards antigens (with multiple repeating epitopes) compared to the monomers. IgM pentamer enables better opsonization of antigens for destruction and fixing complement.

Physical properties

Antibody format: Antiserum

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Reconstitution

Reconstitute with 1.0 mL deionized water (or equivalent).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum Immunoglobulin M Concentration Is Positively Related to Metabolic Syndrome in an Adult Population: Tianjin Chronic Low-Grade Systemic Inflammation and Health (TCLSIH) Cohort Study
Kun Song
PLoS ONE, 9(2), e88701-e88701 (2014)
Structure and function of immunoglobulins.
Schroeder Jr H W and Cavacini L
The Journal of Allergy and Clinical Immunology, 125(2), S41-S52 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service