Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501272

Sigma-Aldrich

Anti-Sin Nombre Virus Glycoprotein 1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$526.00

$526.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 μG
$526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$526.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

virus (Sin Nombre virus)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Sin Nombre virus ... SNVsMgp1(2654026)

General description

Sin Nombre virus (SNV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS). Hantavirus glycoprotein precursor (GPC) is posttranslationally cleaved into two glycoproteins G1 (Gn) and G2 (Gc). The G1 glycoprotein is is thought to be degraded by the host autophagy machinery, and this autophagic clearance is required for efficient virus repliaction.

Immunogen

Sin Nombre virus glycoprotein 1 antibody was raised against a 19 amino acid peptide near the amino terminus of the Sin Nombre virus glycoprotein.The immunogen is located within amino acids 60 - 110 of Sin Nombre Virus Glycoprotein 1.

Application

Sin Nombre virus glycoprotein 1 antibody can detect 10ng Sin Nombre virus glycoprotein 1 peptide in ELISA at 1 μg/ml.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service