Skip to Content
Merck
All Photos(3)

Key Documents

SAB3501142

Sigma-Aldrich

Anti-SARS-CoV-2 (COVID-19) NSP13 (Helicase) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$526.00

$526.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
100 μG
$526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$526.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

SARS coronavirus

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

SARS coronavirus ... ORF1ab(43740578)

Looking for similar products? Visit Product Comparison Guide

General description

Coronavirus disease 2019 (COVID-19), formerly known as 2019-nCoV acute respiratory disease, is an infectious disease caused by SARS-CoV-2, a virus closely related to the SARS virus. The disease is the cause of the 2019–20 coronavirus outbreak. The structure of 2019-nCoV consists of the following: a spike protein (S), hemagglutinin-esterease dimer (HE), a membrane glycoprotein (M), an envelope protein (E) a nucleoclapid protein (N) and RNA. NSP13 is a multi-functional protein with a zinc-binding domain in N-terminus displaying RNA and DNA duplex-unwinding activities with 5′ to 3′ polarity. Activity of Helicase is dependent on magnesium(3,4).

Immunogen

Anti-SARS-CoV-2 (COVID-19) NSP13 (Helicase) antibody was raised against a peptide corresponding to 19 amino acids near the carboxyl terminus of the SARS-CoV-2 (COVID-19) NSP13 (Helicase) protein. The immunogen is located in the last 50 amino acids of SARS-CoV-2 (COVID-19) NSP13 (Helicase).

Application

Antibody validated: Immunohistochemistry in human samples. SARS-CoV-2 (COVID-19) NSP13 antibody can detect 2 ng of free peptide at 1 μg/mL in ELISA. It can detect SARS-CoV-2 NSP13 recombinant protein by ELISA. All other applications and species not yet tested

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service