Skip to Content
Merck
All Photos(1)

Documents

SAB3501027

Sigma-Aldrich

Anti-TRPV1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 92 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPV1(7442)

Immunogen

Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human TRPV1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501027.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua R Smith et al.
Experimental physiology, 105(5), 809-818 (2020-02-28)
What is the central question of this study? How do locomotor muscle metabo- and mechanoreceptor expression compare in heart failure patients and controls? Do relationships exist between the protein expression and cardiopulmonary responses during exercise with locomotor muscle neural afferent
Anna-Lena Klauser et al.
Breast cancer (Dove Medical Press), 13, 659-673 (2021-12-18)
In 2020, breast cancer still represents the most common type of cancer in women worldwide. Depending on the specific molecular subtype, clinical breast cancer management comprises surgery, radiotherapy, chemotherapy and targeted therapy. Furthermore, there are some therapeutic approaches from the
Yuan-Hong Jiang et al.
International journal of molecular sciences, 24(1) (2023-01-09)
The aim of this study was to investigate the expression levels of sensory receptors, inflammatory proteins, and pro-apoptotic proteins in the urothelium of non-Hunner's interstitial cystitis (NHIC) bladders of patients with different clinical and cystoscopic phenotypes. The urothelia from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service