Skip to Content
Merck
All Photos(3)

Documents

SAB3500265

Sigma-Aldrich

Anti-GSTP1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Glutathione S-transferase pi

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTP1(2950)

Immunogen

GSTP1 antibody was raised against a 14 amino acid peptide from near the center of human GSTP1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500265.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio Fuentes-Hernández et al.
Toxicology and applied pharmacology, 378, 114611-114611 (2019-06-10)
Hepatocellular carcinoma (HCC) arises after a long period of exposition to etiological factors that might be either independent or collectively contributing. Several rodent models resemble human HCC; however, the major limitation of these models is the lack of chronic injury
Chao Wu et al.
Experimental and therapeutic medicine, 16(2), 1052-1060 (2018-08-17)
Compound Astragalus and Salvia miltiorrhiza extract (CASE) is a Chinese herbal formula consisting of astragalosides, astragalus polysaccharide and salvianolic acids extracted from Astragalus membranaceus and Salvia miltiorhiza. Previous studies by our group have demonstrated that CASE effectively suppresses diethylinitrosamine (DEN)-induced
Manjeet Deshmukh et al.
Nanomedicine : nanotechnology, biology, and medicine, 14(2), 317-325 (2017-11-22)
Safety is prerequisite for preventive medicine, but non-toxic agents are generally ineffective as clinical chemoprevention. Here we propose a strategy overcoming this challenge by delivering molecular-targeted agent specifically to the effector cell type to achieve sufficient potency, while circumventing toxicity
Brisa Rodope Alarcón-Sánchez et al.
Basic & clinical pharmacology & toxicology, 127(5), 389-404 (2020-06-12)
Alcoholic liver disease (ALD) may be attributed to multiple hits driving several alterations. The aim of this work was to determine whether nucleoredoxin (NXN) interacts with flightless-I (FLII)/actin complex and how this ternary complex is altered during ALD progression induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service