Skip to Content
Merck
All Photos(4)

Key Documents

SAB2701977

Sigma-Aldrich

Anti-DBH antibody produced in rabbit

Synonym(s):

DBM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

69 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:1,000
western blot: 1:500-1:3,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DBH(1621)

General description

Dopamine-β-hydroxylase (DBH) is found in the nuclei of noradrenergic and dopaminergic neurons. The DBH gene is mapped on the human chromosome at 9q34.2.[1]

Immunogen

Recombinant fragment corresponding to a region within amino acids 8 and 320 of Dopamine beta Hydroxylase (Uniprot ID#P09172)

Application

Anti-DBH antibody produced in rabbit has been used in immunohistochemistry (1:200)[2] (1:1000)[3].

Biochem/physiol Actions

Dopamine-β-hydroxylase (DBH) protein plays a role in the conversion of dopamine to noradrenaline. This protein is implicated in various disorders such as Huntington′s chorea, hypertension, Parkinson′s, and smoking disorders. DBH protein is also implicated in heart failure and mental disorders like depression, Tourette syndrome, and schizophrenia. Lower levels of DHB plasma are seen in patients with bipolar disorders and unipolar psychotic depression. Higher levels of DBH plasma are observed in patients with severe depression.[1]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuang Zhang et al.
Journal of neuroscience research, 94(10), 896-906 (2016-06-19)
Rett syndrome (RTT) is a neurodevelopmental disorder affecting multiple functions, including the norepinephrine (NE) system. In the CNS, NE is produced mostly by neurons in the locus coeruleus (LC), where defects in intrinsic neuronal properties, NE biosynthetic enzymes, neuronal CO2
Yao Liu et al.
Experimental physiology, 102(8), 974-984 (2017-06-08)
What is the central question of this study? We investigated the effects of oestrogen and Cimicifuga racemosa on the stellate ganglion, cardiac noradrenaline pathway and Ca2+ -calmodulin-dependent protein kinase II in ovariectomized rats. What is the main finding and its
Marci F Rosenberg et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 43(50), 8621-8636 (2023-10-17)
Astrocytes play essential roles in the developing nervous system, including supporting synapse function. These astrocyte support functions emerge coincident with brain maturation and may be tailored in a region-specific manner. For example, gray matter astrocytes have elaborate synapse-associated processes and
U W Preuss et al.
Drug and alcohol dependence, 133(2), 459-467 (2013-08-03)
Dopamine-beta-hydroxylase (DBH) metabolizes the conversion of dopamine to noradrenaline. DBH, located on chromosome 9q34.2 has variants with potential functional consequences which may be related to alterations of neurotransmitter function and several psychiatric phenotypes, including alcohol dependence (AD), depression (MD) and
Elizabeth S Bucher et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(7), 1128-1137 (2014-04-10)
Neurovascular coupling is understood to be the underlying mechanism of functional hyperemia, but the actions of the neurotransmitters involved are not well characterized. Here we investigate the local role of the neurotransmitter norepinephrine in the ventral bed nucleus of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service