Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501252

Sigma-Aldrich

Anti-CLEC4C antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BDCA2, Anti-CD303, Anti-CLECSF11, Anti-CLECSF7, C-type lectin domain family 4, member C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLEC4C(170482)

General description

C-type lectin domain family 4 member C (CLEC4C) is also known as blood dendritic cell antigen 2. It is expressed on plasmacytoid dendritic cells. CLEC4C is a type II transmembrane protein belonging to the C-type lectin (CTLs) superfamily. The protein consists of an extracellular carbohydrate recognition domain (CRD), a transmembrane region and a short cytoplasmic domain. The gene encoding CLEC4C is localized on human chromosome 12.

Immunogen

Peptide with sequence QSWTKSQKNCSV, from the Internal region of the protein sequence according to NP_569708.1; NP_987099.1.

Biochem/physiol Actions

C-type lectin domain family 4 member C (CLEC4C) is involved in intracellular signal transduction through the transmembrane adaptor, FcεRγ. It inhibits the secretion of soluble TNF-related apoptosis-inducing ligand (TRAIL). CLEC4C may have a role in ligand internalization.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Mechanism for Binding of Galactose-terminated Glycans by the C-type Carbohydrate Recognition Domain in Blood Dendritic Cell Antigen 2.
Jegouzo SA
The Journal of Biological Chemistry, 290(27), 16759-16771 (2015)
Human C-type lectin domain family 4, member C (CLEC4C/BDCA-2/CD303) is a receptor for asialo-galactosyl-oligosaccharides.
Riboldi E
The Journal of Biological Chemistry, 35329-35333 (2011)
BDCA-2, a novel plasmacytoid dendritic cell-specific type II C-type lectin, mediates antigen capture and is a potent inhibitor of interferon alpha/beta induction.
Dzionek A
The Journal of Experimental Medicine, 194(12), 1823-1834 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service