Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501170

Sigma-Aldrich

Anti-Trem2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Trem2a, Anti-Trem2b, Anti-Trem2c, triggering receptor expressed on myeloid cells 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Trem2(83433)

General description

TREM2 (triggering receptor expressed on myeloid cells 2) is a transmembrane receptor belonging to the immunoglobulin (Ig) family. It is composed of an extracellular region containing the Ig-like domain and a cytoplasmic tail. It has a transmembrane region of 33 amino acids followed by a cytoplasmic tail. It is expressed on myeloid cells such as, macrophages found in tissue and dendritic cells, on microglial cells in the brain.

Immunogen

Peptide with sequence CQVEHSTSRNQET, from the internal region of the protein sequence according to NP_112544.1.

Application

Anti-Trem2 antibody produced in goat is suitable for indirect ELISA and western blot assays.

Biochem/physiol Actions

TREM2 (triggering receptor expressed on myeloid cells 2) is involved in the bacterial phagocytosis and regulation of inflammatory response. Its presence in microglia helps to remove the neural debris i.e. amyloid β peptide (Aβ) on microglia. TREM2 suppresses the ability of microglia to phagocytize Aβ, and disturbs the pro-inflammatory activity of these cells. Parkinson′s disease (PD) is portrayed by the degeneration of dopaminergic neurons, in the substantia nigra standards compacta district of the cerebrum. TREM2 binds specifically to the ligands expressed by these dopaminergic neurons. Likewise, microgliosis is additionally involved in the movement of PD. Hence, several type of TREM2 gene might be linked to susceptibility to PD. p.R47H polymorphism in this gene is identified with increased risk of sporadic amyotrophic sidelong sclerosis. Mutation in TREM2 is also associated with polycystic lipomembranous osteodysplaysia with sclerosing leukoencephalop.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linlin Zhang et al.
Frontiers in pharmacology, 13, 894963-894963 (2022-06-21)
Chronic pain after bone fracture and orthopedic surgery is often refractory to most analgesics currently in use, thus emphasizing the urgent need for improved therapeutic medications. Chemokine-dependent neuroinflammation is critical for excitatory synaptic plasticity and central nociception sensitization. Recent studies
Rui-Yuan Pan et al.
Science advances, 5(2), eaau6328-eaau6328 (2019-03-02)
The accumulation of aggregated amyloid-β (Aβ) in the brain is the first critical step in the pathogenesis of Alzheimer's disease (AD), which also includes synaptic impairment, neuroinflammation, neuronal loss, and eventual cognitive defects. Emerging evidence suggests that impairment of Aβ
TREM2 and neurodegenerative disease.
Bruno A Benitez et al.
The New England journal of medicine, 369(16), 1567-1568 (2013-10-18)
Zainularifeen Abduljaleel et al.
PloS one, 9(3), e92648-e92648 (2014-03-26)
Alzheimer's disease is one of the main causes of dementia among elderly individuals and leads to the neurodegeneration of different areas of the brain, resulting in memory impairments and loss of cognitive functions. Recently, a rare variant that is associated
Janet Cady et al.
JAMA neurology, 71(4), 449-453 (2014-02-19)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease in which microglia play a significant and active role. Recently, a rare missense variant (p.R47H) in the microglial activating gene TREM2 was found to increase the risk of several neurodegenerative diseases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service