Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500934

Sigma-Aldrich

Anti-SFRP2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FRP-2, Anti-SARP1, Anti-SDF-5, Anti-Secreted apoptosis related protein 1, Anti-Secreted frizzled-related protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, human, mouse, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFRP2(6423)

Immunogen

Peptide with sequence C-KRWQKGQREFKR from the C Terminus of the protein sequence according to NP_003004.1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Can Xiao et al.
Molecular medicine reports, 10(5), 2293-2298 (2014-09-06)
The role of epigenetic inactivation of secreted frizzled‑related protein 2 (SFRP2) and its functions in the development of oral squamous cell carcinoma (OSCC) remain to be elucidated. The present study demonstrated that SFRP2 mRNA was detected in 97.96% of tumor‑adjacent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service