ADORA2B (adenosine A2b receptor) is a G-protein coupled receptor. It is the most adenosine insensitive adenosine receptor. This gene is located on human chromosome 17p12-p11.2.
Immunogen
Peptide with sequence CQADVKSGNGQ from the C Terminus of the protein sequence according to NP_000667.1.
Biochem/physiol Actions
ADORA2B (adenosine A2b receptor) acts as a therapeutic target for cancer treatment. The tumor cells can use A2b receptor to induce progression. ADORA2B controls immune cell activity. It plays a major role in the transcriptional induction and protein stability of protein period 2 (Per2).
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
New immune checkpoints are emerging in a bid to improve response rates to immunotherapeutic drugs. The adenosine A2A receptor (A2AR) has been proposed as a target for immunotherapeutic development due to its participation in immunosuppression of the tumor microenvironment. Blockade
Journal of the American Society of Nephrology : JASN, 25(3), 547-563 (2013-11-23)
Nucleotide phosphohydrolysis by the ecto-5'-nucleotidase (CD73) is the main source for extracellular generation of adenosine. Extracellular adenosine subsequently signals through four distinct adenosine A receptors (Adora1, Adora2a, Adora2b, or Adora3). Here, we hypothesized a functional role for CD73-dependent generation and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.