Skip to Content
Merck
All Photos(4)

Key Documents

SAB2108302

Sigma-Aldrich

Anti-HNF1A antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

67kDa

species reactivity

human, dog, mouse, guinea pig, horse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNF1A(6927)

General description

Hepatocyte nuclear factor 1α (HNF1α) is a transcription factor that belongs to the liver enriched transcription factor family. It has a N-terminal dimerization domain (amino acids 1–32), a POU-homeobox DNA binding domain (amino acids 150-280) and a C-terminal transactivation domain (amino acids 281-631). HNF1A is located on human chromosome 12q24.

Immunogen

Synthetic peptide directed towards the N terminal region of human HNF1A

Biochem/physiol Actions

HNF1A is required for the expression of several liver specific genes. HNF1A binds to the inverted palindrome 5′-GTTAATNATTAAC-3′.
Hepatocyte nuclear factor 1α (HNF1α) modulates hepatocyte functions. It plays an important role in the modulation of gene expression and replication of hepatitis B virus (HBV). HNF1α is involved in lipid metabolism, gluconeogenesis and deoxygenation of xenobiotics. It stimulates the expression of the preS1 mRNA to induce the production of LHBs (viral large surface protein). Mutations in hepatocyte nuclear factor-1A (HNF1A) results in maturity-onset diabetes of the young type 3 (MODY3).

Sequence

Synthetic peptide located within the following region: HAGQGGLIEEPTGDELPTKKGRRNRFKWGPASQQILFQAYERQKNPSKEE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The HNF1A mutant Ala180Val: Clinical challenges in determining causality of a rare HNF1A variant in familial diabetes.
Sagen JV, et al.
Diabetes Research and Clinical Practice, 133, 142-149 (2017)
p. Q511L mutation of HNF1? in hepatocellular carcinoma suppresses the transcriptional activity and the anti-tumor effect of HNF1α.
Ding CH, et al.
Biochemical and Biophysical Research Communications, 495(1), 86-91 (2018)
Hepatocyte nuclear factor 1α downregulates HBV gene expression and replication by activating the NF-κB signaling pathway.
Lin J, et al.
PLoS ONE, 12(3), e0174017-e0174017 (2017)
Forty-three loci associated with plasma lipoprotein size, concentration, and cholesterol content in genome-wide analysis.
Chasman DI, et al.
PLoS Genetics, 5(11), e1000730-e1000730 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service