Skip to Content
Merck
All Photos(4)

Documents

SAB2103200

Sigma-Aldrich

Anti-GATM antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AGAT, Anti-AT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

dog, rat, guinea pig, horse, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GATM(2628)

Immunogen

Synthetic peptide directed towards the middle region of human GATM

Biochem/physiol Actions

GATM is a mitochondrial enzyme that belongs to the amidinotransferase family. This enzyme is involved in creatine biosynthesis, whereby it catalyzes the transfer of a guanido group from L-arginine to glycine, resulting in guanidinoacetic acid, the immediate precursor of creatine. Mutations in this gene cause arginine:glycine amidinotransferase deficiency, an inborn error of creatine synthesis characterized by mental retardation, language impairment, and behavioral disorders.This gene encodes a mitochondrial enzyme that belongs to the amidinotransferase family. This enzyme is involved in creatine biosynthesis, whereby it catalyzes the transfer of a guanido group from L-arginine to glycine, resulting in guanidinoacetic acid, the immediate precursor of creatine. Mutations in this gene cause arginine:glycine amidinotransferase deficiency, an inborn error of creatine synthesis characterized by mental retardation, language impairment, and behavioral disorders. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: PCFDAADFIRAGRDIFAQRSQVTNYLGIEWMRRHLAPDYRVHIISFKDPN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Wang et al.
Neurological research, 36(3), 224-233 (2014-02-12)
The herbal extract 3-n-butylphthalide (NBP) is used in clinical practice for ischemic patients in China. It has been shown to have various neuroprotective effects both in vitro and in vivo. In the present study, the effects of NBP on learning
Caro-Lyne DesRoches et al.
Human mutation, 37(9), 926-932 (2016-05-29)
Arginine-glycine amidinotransferase (GATM) deficiency is an autosomal-recessive disorder caused by pathogenic variants in GATM. Clinical features include intellectual disability, hypotonia, and myopathy. Due to normal neurodevelopment in asymptomatic individuals on creatine monotherapy, GATM deficiency is a good candidate for newborn
Mike O Becker et al.
American journal of respiratory and critical care medicine, 190(7), 808-817 (2014-09-03)
Systemic sclerosis (SSc)-associated pulmonary arterial hypertension (PAH) portends worse outcome than other forms of PAH. Vasoconstrictive and vascular remodeling actions of endothelin (ET) 1 and angiotensin (Ang) II via endothelin receptor type A (ETAR) and Ang receptor type-1 (AT1R) activation
Jasmine A Luzum et al.
Cell metabolism, 21(4), 622-627 (2015-04-12)
Statin-induced myopathy (SIM) is the most common reason for discontinuation of statin therapy. A polymorphism affecting the gene encoding glycine amidinotransferase (GATM rs9806699 G > A) was previously associated with reduced risk for SIM. Our objective was to replicate the
Marcus E Kleber et al.
Circulation. Cardiovascular genetics, 6(5), 505-513 (2013-09-21)
Low serum levels of the amino acid derivative, homoarginine, have been associated with increased risk of total and cardiovascular mortality. Homoarginine deficiency may be related to renal and heart diseases, but the pathophysiologic role of homoarginine and the genetic regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service