Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102729

Sigma-Aldrich

Anti-XRN1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-5′-3′ exoribonuclease 1, Anti-DKFZp434P0721, Anti-DKFZp686B22225, Anti-DKFZp686F19113, Anti-FLJ41903

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

194 kDa

species reactivity

human, rabbit, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XRN1(54464)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Synthetic peptide directed towards the middle region of human XRN1

Biochem/physiol Actions

SEP1 (XRN1) localizes to cytoplasmic foci containing a complex of mRNA-degrading enzymes. In addition to mRNA metabolism, yeast Sep1 has been implicated in a variety of nuclear and cytoplasmic functions, including homologous recombination, meiosis, telomere maintenance, and microtubule assembly.SEP1 localizes to cytoplasmic foci containing a complex of mRNA-degrading enzymes. In addition to mRNA metabolism, yeast Sep1 has been implicated in a variety of nuclear and cytoplasmic functions, including homologous recombination, meiosis, telomere maintenance, and microtubule assembly.[supplied by OMIM]. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-67 BX640905.1 1-67 68-5188 AY137776.1 1-5121 5189-5269 BG197321.1 308-388 5270-10092 AC023508.16 31718-36540 c

Sequence

Synthetic peptide located within the following region: LPQEISQVNQHHKSGFNDNSVKYQQRKHDPHRKFKEECKSPKAECWSQKM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service