Skip to Content
Merck
All Photos(1)

Documents

SAB2100069

Sigma-Aldrich

Anti-AFF3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AF4/FMR2 family, member 3, Anti-LAF4, Anti-MLLT2-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

133 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AFF3(3899)

General description

AF4/FMR2 family member 3 (AFF3), a tissue-restricted nuclear transcriptional activator, is mainly expressed in the lymphoid tissue. This RNA binding protein is a member of the AF4/LAF4/FMR2/AF5q31 family. AFF3 is a component of the transcriptional super elongation complex. It is expressed at a high level in the thymus. AFF3 has two domains with transactivation activity. This lymphoid-specific gene, is located on the human chromosome 2q11.2.

Immunogen

Synthetic peptide directed towards the middle region of human AFF3

Biochem/physiol Actions

AF4/FMR2 family member 3 (AFF3) participates in transcriptional regulation. It is a potential clinical modulator of estrogen receptor (ER) signaling. This transcription activator can play a role in lymphoid development and cancer. It can bind to double-stranded DNA in vitro. AFF3 overexpression causes tamoxifen resistance and estrogen-independent proliferation in ER+ human breast tumors.

Sequence

Synthetic peptide located within the following region: APATKDSESAPPSHTSDTPAEKALPKSKRKRKCDNEDDYREIKKSQGEKD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yawei Shi et al.
Journal of experimental & clinical cancer research : CR, 37(1), 254-254 (2018-10-18)
Although tamoxifen is a highly effective drug for treating estrogen receptor-positive (ER+) breast cancer, nearly all patients with metastasis with initially responsive tumors eventually relapse, and die from acquired drug resistance. Unfortunately, few molecular mediators of tamoxifen resistance have been
AFF3 (lymphoid nuclear protein related to AF4)
Bergh ARM Von
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2003)
A Ruyssen-Witrand et al.
Tissue antigens, 80(2), 105-118 (2012-07-28)
Rheumatoid arthritis (RA) is a common autoimmune disease with a strong genetic component. Numerous aberrant immune responses have been described during the evolution of the disease. In later years, the appearance of anti-citrullinated protein antibodies (ACPAs) has become a hallmark
Norine Voisin et al.
American journal of human genetics, 108(5), 857-873 (2021-05-08)
The ALF transcription factor paralogs, AFF1, AFF2, AFF3, and AFF4, are components of the transcriptional super elongation complex that regulates expression of genes involved in neurogenesis and development. We describe an autosomal dominant disorder associated with de novo missense variants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service