Skip to Content
Merck
All Photos(4)

Key Documents

SAB1412228

Sigma-Aldrich

ANTI-TSC22D4 antibody produced in mouse

clone 1C12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

THG-1, THG1, TSC22D4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C12, monoclonal

form

buffered aqueous solution

mol wt

antigen 68.97 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSC22D4(81628)

General description

TGF-β-stimulated clone-22 (TSC22) domain family member 4 (TSC22D4) is encoded by the gene mapped to human chromosome 7q22.1. The encoded protein belongs to the TSC22 protein family.
TSC22D4 is a member of the TSC22 domain family of leucine zipper transcriptional regulators (see TSC22D3; MIM 300506) (Kester et al., 1999 [PubMed 10488076]; Fiorenza et al., 2001 [PubMed 11707329]).[supplied by OMIM

Immunogen

TSC22D4 (AAH01966, 1 a.a. ~ 395 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSGGKKKSSFQITSVTTDYEGPGSPGASDPPTPQPPTGPPPRLPNGEPSPDPGGKGTPRNGSPPPGAPSSRFRVVKLPHGLGEPYRRGRWTCVDVYERDLEPHSFGGLLEGIRGASGGAGGRSLDSRLELASLGLGAPTPPSGLSQGPTSWLRPPPTSPGPQARSFTGGLGQLVVPSKAKAEKPPLSASSPQQRPPEPETGESAGTSRAATPLPSLRVEAEAGGSGARTPPLSRRKAVDMRLRMELGAPEEMGQVPPLDSRPSSPALYFTHDASLVHKSPDPFGAVAAQKFSLAHSMLAISGHLDSDDDSGSGSLVGIDNKIEQAMDLVKSHLMFAVREEVEVLKEQIRELAERNAALEQENGLLRALASPEQLAQLPSSGVPRLGPPAPNGPSV

Biochem/physiol Actions

TGF-β-stimulated clone-22 (TSC22) acts as a tumor suppressor. It plays a vital role in systemic glucose metabolism. TSC22D4 inhibition might be an insulin sensitizing route in diabetes treatment. Elevated expression of the gene has been observed in non-small cell lung cancer (NSCLC) patients. Aberrations in the expression of the gene resulted in significant metabolic effects.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun Zhu et al.
Molecular medicine reports, 4(1), 137-143 (2011-04-05)
To identify and provide a global assessment of DNA methylation in fetal ventricular septal defect (VSD), genomic DNA extracted from fetal myocardial tissue samples with VSD (n=21) and from normal fetal myocardial tissue samples (n=15) was analyzed for gene methylation
Julia Jäger et al.
Molecular metabolism, 3(2), 155-166 (2014-03-19)
Disturbances in lipid homeostasis are hallmarks of severe metabolic disorders and their long-term complications, including obesity, diabetes, and atherosclerosis. Whereas elevation of triglyceride (TG)-rich very-low-density lipoproteins (VLDL) has been identified as a risk factor for cardiovascular complications, high-density lipoprotein (HDL)-associated
H A Kester et al.
The Journal of biological chemistry, 274(39), 27439-27447 (1999-09-17)
TGF-beta-stimulated clone-22 (TSC-22) encodes a leucine zipper-containing protein that is highly conserved during evolution. Two homologues are known that share a similar leucine zipper domain and another conserved domain (designated the TSC box). Only limited data are available on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service