Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410883

Sigma-Aldrich

Anti-DHH antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HHG-3, MGC35145

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DHH(50846)

General description

This gene encodes a member of the Hedgehog family. The hedgehog gene family encodes signaling molecules that play an important role in regulating morphogenesis. This protein is predicted to be made as a precursor that is autocatalytically cleaved; the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the organism. Defects in this protein have been associated with partial gonadal dysgenesis (PGD) accompanied by minifascicular polyneuropathy. This protein may be involved in both male gonadal differentiation and perineurial development. (provided by RefSeq)

Immunogen

DHH (NP_066382.1, 1 a.a. ~ 396 a.a) full-length human protein.

Sequence
MALLTNLLPLCCLALLALPAQSCGPGRGPVGRRRYARKQLVPLLYKQFVPGVPERTLGASGPAEGRVARGSERFRDLVPNYNPDIIFKDEENSGADRLMTERCKERVNALAIAVMNMWPGVRLRVTEGWDEDGHHAQDSLHYEGRALDITTSDRDRNKYGLLARLAVEAGFDWVYYESRNHVHVSVKADNSLAVRAGGCFPGNATVRLWSGERKGLRELHRGDWVLAADASGRVVPTPVLLFLDRDLQRRASFVAVETEWPPRKLLLTPWHLVFAARGPAPAPGDFAPVFARRLRAGDSVLAPGGDALRPARVARVAREEAVGVFAPLTAHGTLLVNDVLASCYAVLESHQWAHRAFAPLRLLHALGALLPGGAVQPTGMHWYSRLLYRLAEELLG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service