Skip to Content
Merck
All Photos(3)

Key Documents

SAB1410343

Sigma-Aldrich

Anti-STX2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

EPIM, EPM, MGC51014, STX2A, STX2B, STX2C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 33.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STX2(2054)

General description

Syntaxin 2 (STX2) is encoded by the gene mapped to human chromosome 12q24.33. The encoded protein is ubiquitously expressed and is mainly localized to plasma membrane. STX2 is a member of the soluble N-ethylmaleimide-sensitive factor activating protein receptor (SNARE) membrane fusion machinery.
The product of this gene belongs to the syntaxin/epimorphin family of proteins. The syntaxins are a large protein family implicated in the targeting and fusion of intracellular transport vesicles. The product of this gene regulates epithelial-mesenchymal interactions and epithelial cell morphogenesis and activation. Alternatively spliced transcript variants encoding different isoforms have been identified. (provided by RefSeq)

Immunogen

STX2 (NP_001971.2, 1 a.a. ~ 287 a.a) full-length human protein.

Sequence
MRDRLPDLTACRKNDDGDTVVVVEKDHFMDDFFHQVEEIRNSIDKITQYVEEVKKNHSIILSAPNPEGKIKEELEDLNKEIKKTANKIRAKLKAIEQSFDQDESGNRTSVDLRIRRTQHSVLSRKFVEAMAEYNEAQTLFRERSKGRIQRQLEITGRTTTDDELEEMLESGKPSIFTSDIISDSQITRQALNEIESRHKDIMKLETSIRELHEMFMDMAMFVETQGEMINNIERNVMNATDYVEHAKEETKKAIKYQSKARRKLMFIIICVIVLLVILGIILATTLS

Biochem/physiol Actions

Syntaxin 2 (STX2) plays a vital role in expression, production and release of tissue plasminogen activator (tPA) protein. In mammalian cells, STX2 facilitates the fusion event required for cytokinesis. Deletion in the STX2 gene might be responsible for defective testicular development.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-Wide Association Study for Circulating Tissue Plasminogen Activator Levels and Functional Follow-Up Implicates Endothelial STXBP5 and STX2Significance.
Huang J, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 34(5), 1093-1101 (2014)
Chromosome 12q24.31-q24.33 deletion causes multiple dysmorphic features and developmental delay: First mosaic patient and overview of the phenotype related to 12q24qter defects
Al-Zahrani J, et al.
Molecular Cytogenetics, 4(1), 9-9 (2011)
Syntaxin 2 and Endobrevin Are Required for the Terminal Step of Cytokinesis in Mammalian Cells
Low SH, et al.
Developmental Cell, 4(5), 753-759 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service