Skip to Content
Merck
All Photos(3)

Key Documents

SAB1406777

Sigma-Aldrich

Anti-SART1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Ara1, HOMS1, MGC2038, SART1259, SNRNP110, Snu66

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90.3 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SART1(9092)

General description

This gene encodes two proteins, the SART1(800) protein expressed in the nucleus of the majority of proliferating cells, and the SART1(259) protein expressed in the cytosol of epithelial cancers. The SART1(259) protein is translated by the mechanism of -1 frameshifting during posttranscriptional regulation; its full-length sequence is not published yet. The two encoded proteins are thought to be involved in the regulation of proliferation. Both proteins have tumor-rejection antigens. The SART1(259) protein possesses tumor epitopes capable of inducing HLA-A2402-restricted cytotoxic T lymphocytes in cancer patients. This SART1(259) antigen may be useful in specific immunotherapy for cancer patients and may serve as a paradigmatic tool for the diagnosis and treatment of patients with atopy. The SART1(259) protein is found to be essential for the recruitment of the tri-snRNP to the pre-spliceosome in the spliceosome assembly pathway. (provided by RefSeq)

Immunogen

SART1 (NP_005137.1, 1 a.a. ~ 800 a.a) full-length human protein.

Sequence
MGSSKKHRGEKEAAGTTAAAGTGGATEQPPRHREHKKHKHRSGGSGGSGGERRKRSRERGGERGSGRRGAEAEARSSTHGRERSQAEPSERRVKREKRDDGYEAAASSKTSSGDASSLSIEETNKLRAKLGLKPLEVNAIKKEAGTKEEPVTADVINPMALRQREELREKLAAAKEKRLLNQKLGKIKTLGEDDPWLDDTAAWIERSRQLQKEKDLAEKRAKLLEEMDQEFGVSTLVEEEFGQRRQDLYSARDLQGLTVEHAIDSFREGETMILTLKDKGVLQEEEDVLVNVNLVDKERAEKNVELRKKKPDYLPYAEDESVDDLAQQKPRSILSKYDEELEGERPHSFRLEQGGTADGLRERELEEIRAKLRLQAQSLSTVGPRLASEYLTPEEMVTFKKTKRRVKKIRKKEKEVVVRADDLLPLGDQTQDGDFGSRLRGRGRRRVSEVEEEKEPVPQPLPSDDTRVENMDISDEEEGGAPPPGSPQVLEEDEAELELQKQLEKGRRLRQLQQLQQLRDSGEKVVEIVKKLESRQRGWEEDEDPERKGAIVFNATSEFCRTLGEIPTYGLAGNREEQEELMDFERDEERSANGGSESDGEENIGWSTVNLDEEKQQQDFSASSTTILDEEPIVNRGLAAALLLCQNKGLLETTVQKVARVKAPNKSLPSAVYCIEDKMAIDDKYSRREEYRGFTQDFKEKDGYKPDVKIEYVDETGRKLTPKEAFRQLSHRFHGKGSGKMKTERRMKKLDEEALLKKMSSSDTPLGTVALLQEKQKAQKTPYIVLSGSGKSMNANTITK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service