Skip to Content
Merck
All Photos(3)

Documents

SAB1405710

Sigma-Aldrich

Anti-DFFA antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DFF-45, DFF1, ICAD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~36.5 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DFFA(1676)

General description

The gene DNA fragmentation factor subunit alpha (DFFA), also known as DNA fragmentation factor 45 (DFF45), is mapped to human chromosome 1p36.2. The encoded protein localizes to the chromatin fraction. It exist in association with DFF40, thereby forming a heterodimer. DFFA is popularly called as DFF45 (DNA fragmentation factor 45).

Immunogen

DFFA (NP_004392.1, 1 a.a. ~ 331 a.a) full-length human protein.

Sequence
MEVTGDAGVPESGEIRTLKPCLLRRNYSREQHGVAASCLEDLRSKACDILAIDKSLTPVTLVLAEDGTIVDDDDYFLCLPSNTKFVALASNEKWAYNNSDGGTAWISQESFDVDETDSGAGLKWKNVARQLKEDLSSIILLSEEDLQMLVDAPCSDLAQELRQSCATVQRLQHTLQQVLDQREEVRQSKQLLQLYLQALEKEGSLLSKQEESKAAFGEEVDAVDTGISRETSSDVALASHILTALREKQAPELSLSSQDLELVTKEDPKALAVALNWDIKKTETVQEACERELALRLQQTQSLHSLRSISASKASPPGDLQNPKRARQDPT

Biochem/physiol Actions

DNA fragmentation factor subunit α (DFFA) is a component of DNA fragmentation factor (DFF). It is mainly associated with apoptosis and genomic stability. Caspase-3-mediated cleavage of DFFA releases DFF40 (DNA fragmentation factor 40) which degrades chromosomal DNA. During menstruation, the apoptosis of endometrium cells is associated with DFFA. Its expression decreases after menopause. Silencing of DFFA increases doxorubicin effects in breast cancer cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MiR-145, a new regulator of the DNA fragmentation factor-45 (DFF45)-mediated apoptotic network.
Zhang J, et al.
Molecular Cancer, 9(1), 211-211 (2010)
H W Yang et al.
Neoplasia (New York, N.Y.), 3(2), 165-169 (2001-06-23)
Recently, loss of heterozygosity (LOH) studies suggest that more than two tumor suppressor genes lie on the short arm of chromosome 1 (1p) in neuroblastoma (NB). To identify candidate tumor suppressor genes in NB, we searched for homozygous deletions in
DFF45 expression in human endometrium is associated with menstrual cycle phases and decreases after menopause.
Banas T, et al.
Gynecologic and Obstetric Investigation, 73, 177-182 (2012)
siRNA-mediated knock-down of DFF45 amplifies doxorubicin therapeutic effects in breast cancer cells.
Bagheri F, et al.
Cellular Oncology (Dordrecht), 36, 515-526 (2013)
Yiannis P Ninios et al.
Apoptosis : an international journal on programmed cell death, 15(2), 128-138 (2009-11-03)
A major hallmark of the terminal stages of apoptosis is the internucleosomal DNA fragmentation. The endonuclease responsible for this type of DNA degradation is the DNA fragmentation factor (DFF). DFF is a complex of the endonuclease DFF40 and its chaperone/inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service