Skip to Content
Merck
All Photos(3)

Key Documents

SAB1405672

Sigma-Aldrich

Anti-CTBP2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~56.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTBP2(1488)

General description

This gene produces alternative transcripts encoding two distinct proteins. One protein is a transcriptional repressor, while the other isoform is a major component of specialized synapses known as synaptic ribbons. Both proteins contain a NAD+ binding domain similar to NAD+-dependent 2-hydroxyacid dehydrogenases. A portion of the 3′ untranslated region was used to map this gene to chromosome 21q21.3; however, it was noted that similar loci elsewhere in the genome are likely. Blast analysis shows that this gene is present on chromosome 10. (provided by RefSeq)

Immunogen

CTBP2 (AAH37900.1, 1 a.a. ~ 513 a.a) full-length human protein.

Sequence
MRPGLPGPTGLCAQTSSRGQKSVLKQKESCGIWQLYHFLSRKQEPRWEPCVSGSSSGDGAVADLADELRGYPALCCTLPVHSYRSWAGIRPQIMNGPLHPRPLVALLDGRDCTVEMPILKDLATVAFCDAQSTQEIHEKVLNEAVGAMMYHTITLTREDLEKFKALRVIVRIGSGYDNVDIKAAGELGIAVCNIPSAAVEETADSTICHILNLYRRNTWLYQALREGTRVQSVEQIREVASGAARIRGETLGLIGFGRTGQAVAVRAKAFGFSVIFYDPYLQDGIERSLGVQRVYTLQDLLYQSDCVSLHCNLNEHNHHLINDFTIKQMRQGAFLVNAARGGLVDEKALAQALKEGRIRRAALDVHESEPFSFAQGPLKDAPNLICTPHTAWYSEQASLEMREAAATEIRRAITGRIPESLRNCVNKEFFVTSAPWSVIDQQAIHPELNGATYRYPPGIVGVAPGGLPAAMEGIIPGGIPVTHNLPTVAHPSQAPSPNQPTKHGDNREHPNEQ

Biochem/physiol Actions

The gene CTBP2 (C-terminal binding protein 2) encodes a protein that functions as a transcriptional co-repressor of several tumor suppressor genes resulting in enhanced cancer cell migration and invasion. Its expression is found to be upregulated in hepatocellular carcinoma (HCC). It may be a potential prognostic marker for post liver resection HCC. It is involved in several types of tumor initiation, progression and response to therapy. It is found to interact with the C-terminal region of adenovirus type 2/5 E1A protein, a region that negatively regulates tumorigenicity and the extent of oncogenic transformation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CtBP2 could promote prostate cancer cell proliferation through c-Myc signaling.
Zhang C
Gene, 546, 73-79 (2014)
Xin Zheng et al.
Oncotarget, 6(6), 3752-3769 (2015-02-18)
C-terminal binding protein 2 (CtBP2) is a transcriptional co-repressor that promotes cancer cell migration and invasion by inhibiting multiple tumor suppressor genes that contribute to cell mobility and adhesion. In this investigation, we showed thatCtBP2 expression was increased significantly in
A novel C-terminal binding protein (CTBP2) is closely related to CTBP1, an adenovirus E1A-binding protein, and maps to human chromosome 21q21.3.
Katsanis N and Fisher EM
Genomics, 47, 294-299 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service