Skip to Content
Merck
All Photos(1)

Key Documents

SAB1405610

Sigma-Aldrich

Anti-RCBTB2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CHC1L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
technique(s):
western blot: 1 μg/mL
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RCBTB2(1102)

Related Categories

General description

This gene encodes a member of the RCC1-related GEF family. The N-terminal half of the encoded amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran. (provided by RefSeq)

Immunogen

RCBTB2 (NP_001259.1, 1 a.a. ~ 551 a.a) full-length human protein.

Sequence
MEEELPLFSGDSGKPVQATLSSLKMLDVGKWPIFSLCSEEELQLIRQACVFGSAGNEVLYTTVNDEIFVLGTNCCGCLGLGDVQSTIEPRRLDSLNGKKIACLSYGSGPHIVLATTEGEVFTWGHNAYSQLGNGTTNHGLVPCHISTNLSNKQVIEVACGSYHSLVLTSDGEVFAWGYNNSGQVGSGSTVNQPIPRRVTGCLQNKVVVTIACGQMCCMAVVDTGEVYVWGYNGNGQLGLGNSGNQPTPCRVAALQGIRVQRVACGYAHTLVLTDEGQVYAWGANSYGQLGTGNKSNQSYPTPVTVEKDRIIEIAACHSTHTSAAKTQGGHVYMWGQCRGQSVILPHLTHFSCTDDVFACFATPAVTWRLLSVEPDDHLTVAESLKREFDNPDTADLKFLVDGKYIYAHKVLLKIRCEHFRSSLEDNEDDIVEMSEFSYPVYRAFLEYLYTDSISLSPEEAVGLLDLATFYRENRLKKLCQQTIKQGICEENAIALLSAAVKYDAQDLEEFCFRFCINHLTVVTQTSGFAEMDHDLLKNFISKASRVGAFKN

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service