Skip to Content
Merck

Skip To

SAB1404792

Monoclonal Anti-RBM6, (C-terminal) antibody produced in mouse

clone 4B3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

3G2, DEF-3, DEF3, DKFZp686B0877, FLJ36517, HLC-11, NY-LU-12, g16

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
4B3, monoclonal
Application:
ELISA (c), ELISA (i), WB
Citations:
3


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

capture ELISA: suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBM6(10180)

General description

Mouse monoclonal antibody raised against a partial recombinant RBM6. RNA binding motif protein 6 (RBM6) is expressed in peripheral blood leukocytes, thymus and lymph nodes. The gene encoding it is localized on human chromosome 3p21.3 and has 21 exons.

Immunogen

RBM6 (NP_005768.1, 1024 a.a. ~ 1123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DSPERKRIKYSRETDSDRKLVDKEDIDTSSKGGCVQQATGWRKGTGLGYGHPGLASSEEAEGRMRGPSVGASGRTSKRQSNETYRDAVRRVMFARYKELD

Biochem/physiol Actions

RNA binding motif protein 6 (RBM6) associates with RNA. It has been linked to lung cancers.

Physical form

Solution in phosphate buffered saline, pH 7.4


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ting-lei Gu et al.
Blood, 110(1), 323-333 (2007-03-16)
Activated tyrosine kinases have been frequently implicated in the pathogenesis of cancer, including acute myeloid leukemia (AML), and are validated targets for therapeutic intervention with small-molecule kinase inhibitors. To identify novel activated tyrosine kinases in AML, we used a discovery
Elias G Bechara et al.
Molecular cell, 52(5), 720-733 (2013-12-18)
RBM5, a regulator of alternative splicing of apoptotic genes, and its highly homologous RBM6 and RBM10 are RNA-binding proteins frequently deleted or mutated in lung cancer. We report that RBM5/6 and RBM10 antagonistically regulate the proliferative capacity of cancer cells
Ke Wang et al.
BMC genomics, 8, 348-348 (2007-10-03)
Transcription-induced chimerism, a mechanism involving the transcription and intergenic splicing of two consecutive genes, has recently been estimated to account for approximately 5% of the human transcriptome. Despite this prevalence, the regulation and function of these fused transcripts remains largely



Questions

Reviews

No rating value

Active Filters