Skip to Content
Merck
All Photos(3)

Key Documents

SAB1404387

Sigma-Aldrich

Anti-β-Synuclein (SNCB) Antibody

mouse monoclonal, 3G6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Monoclonal Anti-SNCB antibody produced in mouse, clone 3G6, purified immunoglobulin, buffered aqueous solution

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~40.85 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNCB(6620)

General description

β-Synuclein (SNCB) is present in the presynaptic neuron terminal. It comprises an imperfect KTKEGV sequence at the N-terminus as well as highly acidic and intrinsically disordered regions. SNCB shares sequence similarity with α-synuclein. The SNCB gene is mapped to human chromosome 5q35.2.

Immunogen

SNCB (AAH02902, 1 a.a. ~ 134 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDVFMKGLSMAKEGVVAAAEKTKQGVTEAAEKTKEGVLYVGSKTREGVVQGVASVAEKTKEQASHLGGAVFSGAGNIAAATGLVKREEFPTDLKPEEVAQEAAEEPLIEPLMEPEGESYEDPPQEEYQEYEPEA

Biochem/physiol Actions

β-Synuclein (SNCB) may delay the α- synuclein (αS) fibril formation and inhibit its aggregation. SNCB and αS interact with lipid vesicles to form a high degree of α-helical structure. SNCB is implicated in Dementia with Lewy bodies (DLB), a neurodegenerative dementia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gina M Moriarty et al.
The Journal of biological chemistry, 292(39), 16368-16379 (2017-07-16)
α-Synuclein (αS) is the primary protein associated with Parkinson's disease, and it undergoes aggregation from its intrinsically disordered monomeric form to a cross-β fibrillar form. The closely related homolog β-synuclein (βS) is essentially fibril-resistant under cytoplasmic physiological conditions. Toxic gain-of-function
Tracey Evans et al.
Brain research, 1681, 1-13 (2017-12-27)
Dementia with Lewy bodies (DLB) is the second most prevalent neurodegenerative dementia, where an accumulation of aggregated fibrillar alpha-synuclein in neurons of limbic and forebrain regions of the brain leads to visual hallucination, cognitive impairment of a fluctuating nature and
Andres Jerez et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 30(12), 1343-1349 (2012-03-01)
Interstitial deletions of chromosome 5q are common in acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS), pointing toward the pathogenic role of this region in disease phenotype and clonal evolution. The higher level of resolution of single-nucleotide polymorphism array (SNP-A)
James W P Brown et al.
Scientific reports, 6, 36010-36010 (2016-11-04)
α-Synuclein is an intrinsically disordered protein that is associated with the pathogenesis of Parkinson's disease through the processes involved in the formation of amyloid fibrils. α and β-synuclein are homologous proteins found at comparable levels in presynaptic terminals but β-synuclein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service