Skip to Content
Merck
All Photos(3)

Key Documents

SAB1404348

Sigma-Aldrich

Monoclonal Anti-S100A9 antibody produced in mouse

clone 4G9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.65 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A9(6280)

General description

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis. (provided by RefSeq)

Immunogen

S100A9 (AAH47681, 1 a.a. ~ 114 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MTCKMSQLERNIETIINTFHQYSVKLGHPDTLNQGEFKELVRKDLQNFLKKENRNEKVIEHIMEDLDTNADKQLSFEEFIMLMARLTWASHEKMHEGDEGPGHHHKPGLGEGTP

Application

Monoclonal Anti-S100A9 antibody produced in mouse is suitable for capture ELISA, indirect ELISA and western blot applications.

Biochem/physiol Actions

S100A9 (S100 calcium binding protein A9) has a pro-inflammatory action in acute and chronic inflammation. In addition to inflammatory action, it exhibits cytokine-like activities for the receptor activation of advanced glycation end products (RAGE) and Toll-like receptor 4-dependent signaling cascades. S100A9 have been reported for neutrophil chemotaxis. It stimulates shedding of L-selectin, up-regulates and activates Mac-1 which finally induces neutrophil adhesion to fibrinogen in vitro.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service