Skip to Content
Merck
All Photos(6)

Key Documents

SAB1404277

Sigma-Aldrich

Monoclonal Anti-PTK9, (C-terminal) antibody produced in mouse

clone 1E2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

A6, MGC23788, MGC41876, PTK9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~45.03 kDa

species reactivity

rat, human

technique(s)

capture ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TWF1(5756)

General description

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1. (provided by RefSeq)

Immunogen

PTK9 (AAH22344, 80 a.a. ~ 252 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSSAKPKGPAGKRGIRRLIRGPAETEATTD

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service