Skip to Content
Merck
All Photos(3)

Key Documents

SAB1403687

Sigma-Aldrich

Monoclonal Anti-COX6B1 antibody produced in mouse

clone 2D3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

COX6B, COXG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~35.57 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COX6B1(1340)

General description

Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIb. Three pseudogenes COX6BP-1, COX6BP-2 and COX6BP-3 have been found on chromosomes 7, 17 and 22q13.1-13.2, respectively. (provided by RefSeq)

Immunogen

COX6B1 (AAH01015, 1 a.a. ~ 86 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAEDMETKIKNYKTAPFDSRFPNQNQTRNCWQNYLDFHRCQKAMTAKGGDISVCEWYQRVYQSLCPTSWVTDWDEQRAEGTFPGKI

Application

Monoclonal Anti-COX6B1 antibody produced in mouse is suitable for capture ELISA, indirect ELISA and western blot applications.

Biochem/physiol Actions

COX6B1 (cytochrome c oxidase subunit VIb polypeptide 1) is involved in the regulation of mitochondrial biogenesis. Itis involved in the transport of electron from reduced cytochrome C to molecular oxygen. In muscles, COX gene consists of striated muscle-specific regulatory motifs such as E-box, CArG, and MEF2 at the proximal promoter regions. The large hydrophobic catalytic subunit acts in the electron transfer whereas the small unit is responsible for regulation and assembly of the complex. In mammals, it exists as a monomer with 13 subunits but in the active state serves as a dimer in vivo. Deficiency of COX6B1 causes mitochondrial diseases such as encephalomyopathy, hydrocephalus and hypertropic cardiomyopathy.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulla Najwa Abdulhag et al.
European journal of human genetics : EJHG, 23(2), 159-164 (2014-05-02)
Isolated cytochrome c oxidase (COX) deficiency is a prevalent cause of mitochondrial disease and is mostly caused by nuclear-encoded mutations in assembly factors while rarely by mutations in structural subunits. We hereby report a case of isolated COX deficiency manifesting
N Lenka et al.
Progress in nucleic acid research and molecular biology, 61, 309-344 (1998-09-30)
Cytochrome c Oxidase (COX) is the terminal component of the bacterial as well as the mitochondrial respiratory chain complex that catalyzes the conversion of redox energy to ATP. In eukaryotes, the oligomeric enzyme is bound to mitochondrial innermembrane with subunits
Valeria Massa et al.
American journal of human genetics, 82(6), 1281-1289 (2008-05-24)
Cytochrome c oxidase (COX) deficiency, one of the most common respiratory-chain defects in humans, has been associated with mutations in either mitochondrial DNA genes or nucleus-encoded proteins that are not part in but promote the biogenesis of COX. Mutations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service