Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403138

Sigma-Aldrich

Monoclonal Anti-SLC7A8, (C-terminal) antibody produced in mouse

clone 3F10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

LAT2, LPI-PC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~33.7 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC7A8(23428)

General description

Solute carrier family 7 member 8 (SLC7A8), also known as L-amino acid transporter-2 (LAT-2), is part of the amino acid transporters family. It is a subunit of the heavy chain of the surface antigen 4F2 (4F2hc). The protein is expressed in the placenta and fetal capillaries. The SLC7A8 gene is localized on human chromosome 14q11.2.

Immunogen

SLC7A8 (NP_036376.2, 467 a.a. ~ 535 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VYWQHKPKCFSDFIELLTLVSQKMCVVVYPEVERGSGTEEANEDMEEQQQPMYQPTPTKDKDVAGQPQP

Biochem/physiol Actions

Solute carrier family 7 member 8 (SLC7A8) is involved in amino acid transport.It has a role in absorption of neutral amino acids in the kidney. Mutations in the gene encoding this protein have been linked to lysinuric protein intolerance.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SLC7A7, encoding a putative permease-related protein, is mutated in patients with lysinuric protein intolerance.
Borsani G
Nature Genetics (1999)
Expression and functional characterisation of System L amino acid transporters in the human term placenta.
Gaccioli F
Reproductive Biology and Endocrinology (2015)
Identification of a membrane protein, LAT-2, that Co-expresses with 4F2 heavy chain, an L-type amino acid transport activity with broad specificity for small and large zwitterionic amino acids.
Pineda M
The Journal of Biological Chemistry (1999)
Reabsorption of neutral amino acids mediated by amino acid transporter LAT2 and TAT1 in the basolateral membrane of proximal tubule.
Park SY
Archives of Pharmacal Research (2005)
Wen Deng et al.
Nature communications, 11(1), 304-304 (2020-01-18)
Biological processes in development and disease are controlled by the abundance, localization and modification of cellular proteins. We have developed versatile tools based on recombinant E3 ubiquitin ligases that are controlled by light or drug induced heterodimerization for nanobody or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service