Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401533

Sigma-Aldrich

Monoclonal Anti-SIX2 antibody produced in mouse

clone 1G11, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIX2(10736)

General description

This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila ′sine oculis′ homeobox protein. The encoded protein is a transcription factor which, like other members of this gene family, may be involved in limb or eye development. (provided by RefSeq)

Immunogen

SIX2 (AAH24033, 1 a.a. ~ 291 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSMLPTFGFTQEQVACVCEVLQQGGNIERLGRFLWSLPACEHLHKNESVLKAKAVVAFHRGNFRELYKILESHQFSPHNHAKLQQLWLKAHYIEAEKLRGRPLGAVGKYRVRRKFPLPRSIWDGEETSYCFKEKSRSVLREWYAHNPYPSPREKRELTEATGLTTTQVSNWFKNRRQRDRAAEAKERENNENSNSNSHNPLNGSGKSVLGSSEDEKTPSGTPDHSSSSPALLLSPPPPGLPSLHSLGHPPGPSAVPVPVPGGGGADPLQHHHGLQDSILNPMSANLVDLGS

Biochem/physiol Actions

SIX2 (SIX homeobox 2) is responsible for the regulation of cartilage growth and differentiation in endochondral skeleton. SIX2 also participates in the craniofacial skeletal muscle formation. It might prevent the abnormal drooping eyelids, by weakening the ability of levator muscle to contract. SIX2 haploinsufficiency is linked with congenital ossicle malformation. Mutation in SIX2 is found to be associated with the development of urinary tract, kidney, anterior cranial base, limb tendon and the formation of pyloric sphincter. Deletion in the gene might cause autosomal dominant frontonasal dysplasia syndrome. Mutation in the gene leads to renal hypodysplasia and also chemotherapy-resistant blastemas.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SIX2 haploinsufficiency causes conductive hearing loss with ptosis in humans.
Guan J
Journal of Human Genetics, 61(11), 917-922 (2016)
A new frontonasal dysplasia syndrome associated with deletion of the SIX2 gene.
Hufnagel RB
American Journal of Medical Genetics, 170A(2), 487-491 (2016)
Differential regulation of mouse and human nephron progenitors by the Six family of transcriptional regulators.
O'Brien LL
Development, 143(4), 595-608 (2016)
Nils O Lindström et al.
Journal of the American Society of Nephrology : JASN, 29(3), 825-840 (2018-02-17)
The nephron is the functional unit of the kidney, but the mechanism of nephron formation during human development is unclear. We conducted a detailed analysis of nephron development in humans and mice by immunolabeling, and we compared human and mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service