Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401484

Sigma-Aldrich

Anti-PREPL antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ16627, KIAA0436

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PREPL(9581)

General description

PREPL belongs to the prolyl oligopeptidase subfamily of serine peptidases (Parvari et al., 2005 [PubMed 15913950]).[supplied by OMIM

Immunogen

PREPL (NP_006027, 1 a.a. ~ 727 a.a) full-length human protein.

Sequence
MQQKTKLFLQALKYSIPHLGKCMQKQHLNHYNFADHCYNRIKLKKYHLTKCLQNKPKISELARNIPSRSFSCKDLQPVKQENEKPLPENMDAFEKVRTKLETQPQEEYEIINVEVKHGGFVYYQEGCCLVRSKDEEADNDNYEVLFNLEELKLDQPFIDCIRVAPDEKYVAAKIRTEDSEASTCVIIKLSDQPVMEASFPNVSSFEWVKDEEDEDVLFYTFQRNLRCHDVYRATFGDNKRNERFYTEKDPSYFVFLYLTKDSRFLTINIMNKTTSEVWLIDGLSPWDPPVLIQKRIHGVLYYVEHRDDELYILTNVGEPTEFKLMRTAADTPAIMNWDLFFTMKRNTKVIDLDMFKDHCVLFLKHSNLLYVNVIGLADDSVRSLKLPPWACGFIMDTNSDPKNCPFQLCSPIRPPKYYTYKFAEGKLFEETGHEDPITKTSRVLRLEAKSKDGKLVPMTVFHKTDSEDLQKKPLLVHVYGAYGMDLKMNFRPERRVLVDDGWILAYCHVRGGGELGLQWHADGRLTKKLNGLADLEACIKTLHGQGFSQPSLTTLTAFSAGGVLAGALCNSNPELVRAVTLEAPFLDVLNTMMDTTLPLTLEELEEWGNPSSDEKHKNYIKRYCPYQNIKPQHYPSIHITAYENDERVPLKGIVSYTEKLKEAIAEHAKDTGEGYQTPNIILDIQPGGNHVIEDSHKKITAQIKFLYEELGLDSTSVFEDLKKYLKF

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-omics analyses identify PREPL as a key regulator of protein trafficking, a pathway underlying Alzheimer's disease pathogenesis.
Duarte, et al.
Molecular Pharmacology, 104, 16-Jan-16-Jan (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service