SAB1401127
Anti-EPS8 antibody produced in mouse
purified immunoglobulin, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... EPS8(2059)
General description
This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. (provided by RefSeq)
Immunogen
EPS8 (NP_004438.3, 1 a.a. ~ 822 a.a) full-length human protein.
Sequence
MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH
Sequence
MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH
Biochem/physiol Actions
EPS8 (epidermal growth factor receptor pathway substrate 8) is a substrate for tyrosine kinases, such as epidermal growth factor receptor (EGFR), fibroblast growth factor receptor (FGFR), platelet-derived growth factor receptor (PDGFR) and ERBB2 (erb-b2 receptor tyrosine kinase 2). In addition, it also interacts with the Src non-receptor tyrosine kinase. The protein is involved in Rac signaling and receptor endocytosis. It also regulates dendritic cell migration, morphogenesis of intestinal cells and microvilli, and stereocilia activity. It is upregulated in pancreatic cancer and oral squamous cell carcinoma. Mutation in EPS8 is associated with autosomal recessive profound deafness.
Physical form
Solution in phosphate buffered saline, pH 7.4
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Orphanet journal of rare diseases, 9, 55-55 (2014-04-20)
Almost 90% of all cases of congenital, non-syndromic, severe to profound inherited deafness display an autosomal recessive mode of transmission (DFNB forms). To date, 47 causal DFNB genes have been identified, but many others remain to be discovered. We report
Journal of cell science, 127(Pt 24), 5303-5316 (2014-11-02)
Eps8 is an actin regulatory scaffold protein whose expression is increased in squamous cell carcinoma (SCC) cells. It forms a complex with both focal adhesion kinase (FAK, also known as PTK2) and Src in SCC cells derived from skin carcinomas
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service