Skip to Content
Merck
All Photos(3)

Key Documents

SAB1400775

Sigma-Aldrich

Anti-CDCA5 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MGC16386

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDCA5(113130)

Related Categories

General description

CDCA5 (cell division cycle associated 5) gene is mapped to human chromosome 11q13.1. The gene codes for the protein sororin. The protein is localized in the nucleus. It is known to be conserved among the vertebrates.

Immunogen

CDCA5 (NP_542399.1, 1 a.a. ~ 252 a.a) full-length human protein.

Sequence
MSGRRTRSGGAAQRSGPRAPSPTKPLRRSQRKSGSELPSILPEIWPKTPSAAAVRKPIVLKRIVAHAVEVPAVQSPRRSPRISFFLEKENEPPGRELTKEDLFKTHSVPATPTSTPVPNPEAESSSKEGELDARDLEMSKKVRRSYSRLETLGSASTSTPGRRSCFGFEGLLGAEDLSGVSPVVCSKLTEVPRVCAKPWAPDMTLPGISPPPEKQKRKKKKMPEILKTELDEWAAAMNAEFEAAEQFDLLVE

Biochem/physiol Actions

Sororin regulates proper cohesion and separation of sister chromatids during replication, by stabilizing cohesin on DNA. Phosphorylation of sororin by cyclin B during mitosis, reduces its potential to prevent Wap1 (cohesin release factor) and Pds5 (cohesin-interacting protein) binding.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomic alterations identified by array comparative genomic hybridization as prognostic markers in tamoxifen-treated estrogen receptor-positive breast cancer.
Han W
BMC Cancer, 6:92, 1-13 (2006)
C-terminus of Sororin interacts with SA2 and regulates sister chromatid cohesion.
Zhang N and Pati D
Cell Cycle, 14(6), 820-826 (2015)
Interaction of Sororin protein with polo-like kinase 1 mediates resolution of chromosomal arm cohesion.
Zhang N.
The Journal of Biological Chemistry, 286(48), 41826-41837 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service