Skip to Content
Merck
All Photos(2)

Documents

SAB1400422

Sigma-Aldrich

Anti-SEC23IP antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MSTP053, Anti-P125

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEC23IP(11196)

General description

COPII-coated vesicles are involved in protein transport from the endoplasmic reticulum to the Golgi apparatus. The protein encoded by this gene was identified by its interaction with a mouse protein similar to yeast Sec23p, an essential component of the COPII. This protein shares significant similarity with phospholipid-modifying proteins, especially phosphatidic acid preferring-phospholipase A1. Overexpression of this protein has been shown to cause disorganization of the endoplasmic reticulum-Golgi intermediate compartment and Golgi apparatus, which suggests its role in the early secretory pathway. (provided by RefSeq)

Immunogen

SEC23IP (N/A, 1 a.a. ~ 486 a.a) full-length human protein.

Sequence
MAERKPNGGSGGASTSSSGTNLLFSSSATEFSFNVPFIPVTQASASPASLLLPGEDSTDVGEEDSFLGQTSIHTSAPQTFSYFSQVSSSSDPFGNIGQSPLTTAATSVGQSGFPKPLTALPFTTGSQDVSNAFSPSISKAQPGAPPSSLMGINSYLPSQPSSLPPSYFGNQPQGIPQPGYNPYRHTPGSSRANPYIAPPQLQQCQTPGPPAHPPPSGPPVQMYQMPPGSLPPVPSSVQSPAQQQVPARPGAPSVQVPSPFLLQNQYEPVQPHWFYCKEVEYKQLWMPFSVFDSLNLEEIYNSVQPDPESVVLGTDGGRYDVYLYDRIRKAAYWEEEPAEVRRCTWFYKGDTDSRFIPYTEEFSEKLEAEYKKAVTTNQWHRRLEFPSGETIVMHNPKVIVQFQPSSVPDEWGTTQDGQTRPRVVKRGIDDNLDEIPDGEMPQVDHLVFVVHGIGPVCDLRFRSIIECVDDLGWFLSNCCGHISRNL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service