Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400082

Sigma-Aldrich

Anti-E4F1 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-E4F, Anti-MGC99614

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... E4F1(1877)

Related Categories

General description

The zinc finger protein encoded by this gene is one of several cellular transcription factors whose DNA-binding activities are regulated through the action of adenovirus E1A. A 50-kDa amino-terminal product is generated from the full-length protein through proteolytic cleavage. The protein is differentially regulated by E1A-induced phosphorylation. The full-length gene product represses transcription from the E4 promoter in the absence of E1A, while the 50-kDa form acts as a transcriptional activator in its presence. (provided by RefSeq)

Immunogen

E4F1 (AAH80524.1, 1 a.a. ~ 784 a.a) full-length human protein.

Sequence
MEGAMAVRVTAAHTAEAQAEAGREAGEGAVAAVAAALAPSGFLGLPAPFSEEDEDDVHRCGRCQAEFTALEDFVQHKIQKACQRAPPEALPATPATTALLGQEVVPAAPGPEEPITVAHIVVEAASLAADISHASDLVGGGHIKEVIVAAEAELGDGEMAEAPGSPHQQGLGLAGEGEQAQVKLLVNKDGRYVCALCHKTFKTGSILKAHMVTHSSRKDHECKLCGASFRTKGSLIRHHRRHTDERPYKCSKCGKSFRESGALTRHLKSLTPCTEKIRFSVSKDVVVSKEDARAGSGAGAAGLGTATSSVTGEPIETSPVIHLVTDAKGTVIHEVHVQMQELSLGMKALAPEPPVSQELPCSSEGSRENLLHQAMQNSGIVLERAAGEEGALEPAPAAGSSPQPLAVAAPQLPVLEVQPLETQVASEASAVPRTHPCPQCSETFPTAATLEAHKRGHTGPRPFACAQCGKAFPKAYLLKKHQEVHVRERRFRCGDCGKLYKTIAHVRGHRRVHSDERPYPCPKCGKRYKTKNAQQVHFRTHLEEKPHVCQFCSRGFREKGSLVRHVRHHTGEKPFKCYKCGRGFAEHGTLNRHLRTKGGCLLEVEELLVSEDSPAAATTVLTEDPHTVLVEFSSVVADTQEYIIEATADDAETSEATEIIEGTQTEVDSHIMKVVQQIVHQASAGHQIIVQNVTMDEETALGPEAAAADTITIATPESLTEQVAMTLASAISEGTVLAARAGTSGTEQATVTMVSSEDIEILEHAGELVIASPEGQLEVQTVIV

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service