Skip to Content
Merck
All Photos(6)

Key Documents

SAB1400018

Sigma-Aldrich

Monoclonal Anti-RHOA antibody produced in mouse

clone 1C1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ARH12, Anti-ARHA, Anti-RHO12, Anti-RHOH12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1λ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOA(387)

Immunogen

RHOA (AAH01360, 1 a.a. ~ 193 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAAIRKKLVIVGDGACGKTCLLIVFSKDQFPEVYVPTVFENYVADIEVDGKQVELALWDTAGQEDYDRLRPLSYPDTDVILMCFSIDSPDSLENIPEKWTPEVKHFCPNVPIILVGNKKDLRNDEHTRRELAKMKQEPVKPEEGRDMANRIGAFGYMECSAKTKDGVREVFEMATRAALQARRGKKKSGCLVL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianjun Liu et al.
The Journal of pathology, 249(1), 126-136 (2019-05-06)
RhoA has been identified as having a gain-of-function mutation in approximately 20% of diffuse gastric cancer patients. However, the carcinogenic role of RhoA mutations in gastric cancer (GC) is unclear. In the present study, we report that RhoA directly interacts
Jianjun Liu et al.
The Journal of pathology, 249(1), 126-136 (2019-05-06)
RhoA has been identified as having a gain-of-function mutation in approximately 20% of diffuse gastric cancer patients. However, the carcinogenic role of RhoA mutations in gastric cancer (GC) is unclear. In the present study, we report that RhoA directly interacts
Seungyoon Nam et al.
BMC cancer, 22(1), 798-798 (2022-07-20)
Gastric cancer (GC) is one of the most fatal cancers worldwide and is generally only detected after it has progressed to an advanced stage. Since there is a lack of comprehensive data on RHOA protein expression of patients with GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service