Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306514

Sigma-Aldrich

ANTI-DCD(C-TERMINAL) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AIDD, DCD, DSEP, Dermcidin, Preproteolysin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

11284 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCD(117159)

Related Categories

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiko Mizukawa et al.
Archives of dermatological research, 311(1), 71-82 (2018-12-05)
No previous studies have convincingly linked sweating disturbance with the subsequent development of lichen planus (LP). Therefore, we investigated whether sweating disturbance could be specifically detected in LP lesions and how it could trigger lesion development. We utilized the impression
Jianfeng Zhang et al.
Oncology reports, 40(5), 2854-2863 (2018-09-19)
Long non‑coding RNAs (lncRNAs) perform distinct biological functions by regulating gene expression through various molecular mechanisms under normal physiological and pathological conditions. However, the function of the stomach cancer‑associated transcript‑3 (STCAT3) lncRNA, including its prognostic significance and role as a
Margaret Coates et al.
PloS one, 14(5), e0216249-e0216249 (2019-05-07)
Hidradenitis suppurativa (HS) is a debilitating chronic inflammatory skin disease resulting in non-healing wounds affecting body areas of high hair follicle and sweat gland density. The pathogenesis of HS is not well understood but appears to involve dysbiosis-driven aberrant activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service