Skip to Content
Merck
All Photos(2)

Key Documents

SAB1300980

Sigma-Aldrich

Anti-DGKA (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

80 kDa diacylglycerol kinase, Anti-DAG kinase α, Anti-DGK-α, Anti-Diacylglycerol kinase α, Anti-Diglyceride kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... DGKA(1606)

General description

Upon cell stimulation, the kinase DGKA converts the second messenger diacylglycerol into phophatidate, initiating the resynthesis of phosphatidylinositols and attenuating protein kinase C activity. DGKA is stimulated by calcium and phosphatidylserine, and is phosphorylated by protein kinase C. Tissue expression is in lymphocytes and oligodengroglial cells. DGKA contains 2 zinc-dependent phorbol-ester and DAG binding domains, and 2 EF-hand calcium binding domains.

Immunogen

DGKA (NP_958853, 603-638)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human DGKA.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service