Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300099

Sigma-Aldrich

Anti-KIST (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-KIS, Anti-Kinase interacting with stathmin, Anti-Serine/threonine-protein kinase Kist, Anti-UHMK1, U2AF homology motif kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIST(127933)

General description

KIST, a member of the Ser/Thr protein kinase family, is a pyruvate kinase that catalyzes formation of phosphoenolpyruvate from pyruvate and ATP. A role for the primarily nuclear KIST protein in mediation of cellular metabolism has been postulated based on the interaction identified with thyroid hormone. KIST is widely expressed, with highest abundance in skeletal muscle, kidney, placenta and peripheral blood leukocytes. Upon serum stimulation, KIST phosphorylates CDKN1B/p27Kip1, thereby regulating the subcellular location of CDKN1B and cell cycle progression in the G1 phase. KIST, which contains one RNA recognition motif domain, has been proposed to partipate in trafficking and processing of RNA. KIST binds to Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells.

Immunogen

KIST (NP_787062, 270-305)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human KIST.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service