Skip to Content
Merck
All Photos(3)

Key Documents

SAB1100176

Sigma-Aldrich

Anti-GUCY2C (981-995) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-GUC2C, Anti-Guanylate cyclase 2C (heat stable enterotoxin receptor), Anti-STAR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~123 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GUCY2C(2984)

General description

The guanylate cyclase 2C (GUCY2C) gene is mapped to human chromosome 12p12. The gene codes for a transmembrane receptor guanylate cyclase C (GC-C) and is expressed in intestine.

Immunogen

synthetic peptide corresponding to amino acids 981-995 of human GUCY2C

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Guanylate cyclase 2C (GUCY2C) converts guanosine-5′-triphosphate (GTP) to cyclic guanosine monophosphate (cGMP), upon binding of its ligand guanylin and uroguanylin, which are paracrine hormones. GUCY2C functions as a tumor suppressor and hinders intestinal tumorigenesis by inhibiting the protein kinase B/AKT signaling pathway. Transmembrane receptor guanylate cyclase C (GC-C) regulates the secretion of chloride. Alteration in the gene expression leads to congenital sodium diarrhoea (CSD).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congenital secretory diarrhoea caused by activating germline mutations in GUCY2C
Muller T, et al.
Gut, 65, 1306-1313 (2019)
The hormone receptor GUCY2C suppresses intestinal tumor formation by inhibiting AKT signaling.
Lin JE
Gastroenterology, 138, 241-254 (2010)
Localization of the guanylyl cyclase C gene to mouse chromosome 6 and human chromosome 12p12.
Mann EA
Genomics, 34, 265-267 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service